Skip to Content
Merck
All Photos(1)

Documents

45436

Supelco

Dichlorprop

PESTANAL®, analytical standard

Synonym(s):

2-(2,4-Dichlorophenoxy)propionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H8Cl2O3
CAS Number:
Molecular Weight:
235.06
Beilstein:
6568822
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

quality

racemate

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CC(Oc1ccc(Cl)cc1Cl)C(O)=O

InChI

1S/C9H8Cl2O3/c1-5(9(12)13)14-8-3-2-6(10)4-7(8)11/h2-5H,1H3,(H,12,13)

InChI key

MZHCENGPTKEIGP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wenbang Li et al.
Molecules (Basel, Switzerland), 24(9) (2019-05-01)
Chlorophenoxy acid herbicides (CAHs), which are widely used on cereal crops, have become an important pollution source in grains. In this work, a highly hydrophobic poly (octadecyl methacrylate-co-trimethylolpropane trimethacrylate) [poly (OMA-co-TRIM)] monolithic column has been specially prepared for hydrophobic in-tube
Hirotatsu Murano et al.
Journal of agricultural and food chemistry, 56(4), 1350-1357 (2008-02-06)
The adsorption of 2-(2,4-dichloro-3-methylphenoxy)propanoic acid (DMPA) on the surface horizon of a humus-rich Andosol was examined. To investigate the mechanisms of adsorption, chemically treated Andosols, such as organic matter removed Andosol, organic matter and active metals removed Andosol, and clay
Yuezhong Wen et al.
Environmental science & technology, 45(11), 4778-4784 (2011-05-07)
Reactive oxygen species (ROS) are considered to be the key players in cell toxicity. However, cross talk between the enantioselective toxicity of pesticides, heavy metals, and ROS is poorly understood. To decipher the puzzle, the effects of copper (Cu) on
Yuezhong Wen et al.
Environmental science & technology, 44(13), 4981-4987 (2010-06-12)
Chitosan, a natural polysaccharide, is widely regarded as a biocompatible and potential carrier for controlled-release drug formulations. However, the potential effect of chitosan on the environmental behavior of contaminants is poorly understood, especially on chiral chemicals. In this study, the
Hui Chen et al.
Aquatic toxicology (Amsterdam, Netherlands), 120-121, 19-26 (2012-05-23)
The effect of heavy metal toxicity on the environment is usually linked to changes in the glutathione redox cycle and oxidative damage as causative events. However, it is unknown whether changes in the glutathione redox cycle are a cause or

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service