Skip to Content
Merck
All Photos(2)

Key Documents

40446

Supelco

Pararot

analytical standard

Synonym(s):

Para Red, 1-(4-Nitrophenylazo)-2-naphthol, 1-(p-Nitrophenylazo)-2-naphthol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H4N=NC10H6OH
CAS Number:
Molecular Weight:
293.28
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥97.0% (HPLC)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

248-252 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

Oc1ccc2ccccc2c1N=Nc3ccc(cc3)[N+]([O-])=O

InChI

1S/C16H11N3O3/c20-15-10-5-11-3-1-2-4-14(11)16(15)18-17-12-6-8-13(9-7-12)19(21)22/h1-10,20H

InChI key

WOTPFVNWMLFMFW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service