Skip to Content
Merck
All Photos(1)

Key Documents

34518

Supelco

Trifloxystrobin Metabolite CGA 321113

PESTANAL®, analytical standard

Synonym(s):

(E)-α-Methoxyimino-2-[(E)-1-(3-trifluoromethylphenyl)ethylidenaminooxymethyl]phenylacetic acid, Trifloxystrobin acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H17F3N2O4
CAS Number:
Molecular Weight:
394.34
UNSPSC Code:
77101502
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

application(s)

agriculture
environmental

format

neat

Related Categories

General description

Trifloxystrobin Metabolite CGA 321113 is an acid metabolite of trifloxystrobin, which belongs to the class of strobilurin broad-spectrum fungicides. Its mode of action involves its binding to the Q0 site of cytochrome b, thereby resulting in the inhibition of mitochondrial respiration of the fungi. Trifloxystrobin Metabolite CGA 321113 is largely utilized for the management of powdery mildew as well as anthracnose, that adversely affect fruits and agriculture products.

Application

Trifloxystrobin Metabolite CGA 321113 may be used as an analytical standard for the determination of the analyte in agricultural products, fruits, and environmental samples by various chromatography techniques.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dissipation kinetics of trifloxystrobin and tebuconazole on apple (Malus domestica) and soil-A multi location study from north western Himalayan region
Patyal SK, et al.
Chemosphere, 92(8), 949-954 (2013)
Determination of Trifloxystrobin and Its Metabolites in Hawaii Soils by ASE- LC-MS/MS
Chen J, et al.
Journal of Agricultural and Food Chemistry, 56(6), 1829-1837 (2008)
Dissipation and residues of trifloxystrobin and its metabolite in rice under field conditions
Li P, et al.
Environmental Toxicology and Chemistry / Setac, 33(12), 2654-2660 (2014)
Simultaneous determination of tebuconazole, trifloxystrobin, and its metabolite trifloxystrobin acid residues in gherkin under field conditions
Paramasivam M, et al.
Journal of Separation Science, 38(6), 958-964 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service