Skip to Content
Merck
All Photos(2)

Key Documents

33442

Supelco

Imiprothrin

PESTANAL®, analytical standard

Synonym(s):

2-Propargylsuccinimidylmethyl chrysanthemate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H22N2O4
CAS Number:
Molecular Weight:
318.37
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

description

mixture of isomers

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

C\C(C)=C/C1C(C(=O)OCN2C(=O)CN(CC#C)C2=O)C1(C)C

InChI

1S/C17H22N2O4/c1-6-7-18-9-13(20)19(16(18)22)10-23-15(21)14-12(8-11(2)3)17(14,4)5/h1,8,12,14H,7,9-10H2,2-5H3

InChI key

VPRAQYXPZIFIOH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Imiprothrin is a kind of pyrethroid consisting of an imidazolydinyl ring, which can find applications as a potent knock-down insecticide.
Imiprothrin, belongs to the class of pyrethroids, having high insecticidal potency. These pesticides are widely used in the field of agriculture, household, forestry, and horticulture, for the control of outdoor insect pests and other household pests.

Application

Imiprothrin may be used as an analytical reference standard for the quantification of the analyte in whole blood, air samples, and indoor insecticide products, river water samples using chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - STOT SE 2

Target Organs

Nervous system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F

Flash Point(C)

110 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aerobic metabolism and adsorption of pyrethroid insecticide imiprothrin in soil
Suzuki Y, et al.
Journal of pest science, 31, 322-328 (2006)
Simultaneous determination of 18 pyrethroids in indoor air by gas chromatography/mass spectrometry
Yoshida T, et al.
Journal of Chromatography A, 1216(26), 5069-5076 (2009)
S Dzib-Florez et al.
Journal of the American Mosquito Control Association, 36(1), 16-21 (2020-06-05)
Commercial aerosolized insecticides can be implemented as a community-based approach to targeted indoor residual spraying against Aedes aegypti, but their efficacy on pyrethroid-resistant mosquitoes has not yet been evaluated. Two commercial aerosolized products (H24 Poder Fulminante Ultra Eficaz®, carbamate, and
Ultrasound-assisted dispersive liquid--liquid microextraction for the determination of six pyrethroids in river water
Yan H, et al.
Journal of Chromatography A, 1217(32), 5152-5157 (2010)
Simultaneous determination of nine pyrethroids in indoor insecticide products by capillary gas chromatography
Yan H, et al.
Journal of Pharmaceutical and Biomedical Analysis, 51(3), 774-777 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service