Skip to Content
Merck
All Photos(3)

Key Documents

291056

Sigma-Aldrich

Methyl formate

anhydrous, 99%

Synonym(s):

Formic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCO2CH3
CAS Number:
Molecular Weight:
60.05
Beilstein:
1734623
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.21
grade:
anhydrous
Assay:
99%
bp:
32-34 °C (lit.)
vapor pressure:
32.91 psi ( 55 °C)
9.47 psi ( 20 °C)

grade

anhydrous

Quality Level

vapor density

2.1 (vs air)

vapor pressure

32.91 psi ( 55 °C)
9.47 psi ( 20 °C)

Assay

99%

form

liquid

autoignition temp.

842 °F

expl. lim.

23 %

impurities

<0.005% water

evapn. residue

<0.0005%

refractive index

n20/D 1.343 (lit.)

pH

4-5 (20 °C, 200 g/L)

bp

32-34 °C (lit.)

mp

−100 °C (lit.)

density

0.974 g/mL at 20 °C (lit.)

SMILES string

[H]C(=O)OC

InChI

1S/C2H4O2/c1-4-2-3/h2H,1H3

InChI key

TZIHFWKZFHZASV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Methyl formate may be used in the synthesis of the following building blocks:
  • methyl 4-chloro-2-formylbutanoate from methyl 4-chlorobutanoate via -formylation
  • potassium methyl α-[(methoxyethylidene)amino]-β-hydroxyacrylate from methyl α-[(methoxyethylidene)amino]acetate
  • (S)-(-)-1-formyl-2-hydroxymethylpyrrolidine from (S)-(+)-2-hydroxymethylpyrrolidine via N-formylation

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Irrit. 2 - Flam. Liq. 1 - STOT SE 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-2.2 °F - closed cup

Flash Point(C)

-19 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

(S)-(-)-1-Amino-2-Methoxymethylpyrrolidine (SAMP) and (R)-(+)-1-Amino-2-Methoxymethylpyrrolidine (RAMP), Versatile Chiral Auxiliaries
Enders D, et al.
Organic Syntheses, 65, 173-173 (1987)
4-Methoxycarbonyl-2-methyl-1,3-oxazole
White JD, et al
Organic Syntheses, 79, 244-244 (2002)
Katherine R Phillips et al.
Journal of the American Chemical Society, 135(2), 574-577 (2012-12-28)
Methyl formate is produced from the photo-oxidation of methanol on preoxidized TiO(2)(110). We demonstrate that two consecutive photo-oxidation steps lead to methyl formate using mass spectrometry and scanning tunneling microscopy. The first step in methanol oxidation is formation of methoxy
Methyl N-Phenyl Carbamate Synthesis from Aniline and Methyl Formate: Carbon Recycling to Chemical Products.
Yalfani MS, et al.
ChemSusChem, 8(3), 443-447 (2015)
Synthesis of Methyl 1-Formylcyclopropanecarboxylate utilizing Ti-Claisen Condensation
Ashida Y, et al.
Organic Syntheses, 93, 286-286 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service