Skip to Content
Merck
All Photos(1)

Documents

23270

Sigma-Aldrich

Chloramine T trihydrate

purum p.a., for the detection of halogens and bromate, ≥98.0% (RT)

Synonym(s):

N-Chloro-p-toluenesulfonamide sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4SO2NClNa · 3H2O
CAS Number:
Molecular Weight:
281.69
Beilstein:
3924168
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

grade

purum p.a.

Quality Level

Assay

≥98.0% (RT)

quality

for the detection of halogens and bromate

reaction suitability

reagent type: oxidant

mp

167-170 °C (lit.)

SMILES string

O.O.O.Cc1ccc(cc1)S(=O)(=O)N([Na])Cl

InChI

1S/C7H7ClNO2S.Na.3H2O/c1-6-2-4-7(5-3-6)12(10,11)9-8;;;;/h2-5H,1H3;;3*1H2/q-1;+1;;;

InChI key

NZYOAGBNMCVQIV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Enhancement of Photocatalytic and Antimicrobial Potential: Research into nanocomposites loaded with Chloramine T trihydrate has shown enhanced photocatalytic and antimicrobial properties, suggesting applications in material sciences and environmental engineering (Elkodous et al., 2020).

Other Notes

Review; Oxidizing agent for the preparation of 131I-labeled material with high specific radioactivity; Reagent for the selective oxidation of methionine

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Resp. Sens. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

377.6 °F - closed cup

Flash Point(C)

192 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Crystal and molecular structure of chloramine-T trihydrate. Absence of a sodium-nitrogen interaction in the oxidant. N-chloro-N-sodiotoluene-p-sulfonamide.
Olmstead MM and Power PP.
Inorganic Chemistry, 25(22), 4057-4058 (1986)
D.H. Bremner
Synthetic Reagents, 6, 9-9 (1985)
THE PREPARATION OF I-131-LABELLED HUMAN GROWTH HORMONE OF HIGH SPECIFIC RADIOACTIVITY.
F C GREENWOOD et al.
The Biochemical journal, 89, 114-123 (1963-10-01)
The estimation of microgram amounts of methionine by reaction with chloramine-T.
G E Trout
Analytical biochemistry, 93(2), 419-422 (1979-03-01)
Chloramine T and related N-halogeno-N-metallo reagents.
Campbell MM and Johnson G.
Chemical Reviews, 78(1), 65-87 (1978)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service