Skip to Content
Merck
All Photos(3)

Key Documents

216208

Sigma-Aldrich

Hydrazine dihydrochloride

≥98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2NH2 · 2HCl
CAS Number:
Molecular Weight:
104.97
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.21
Assay:
≥98%
form:
powder, crystals or chunks
solid

Quality Level

Assay

≥98%

form

powder, crystals or chunks
solid

mp

200 °C (dec.) (lit.)

density

1.42 g/mL at 25 °C (lit.)

SMILES string

Cl[H].Cl[H].NN

InChI

1S/2ClH.H4N2/c;;1-2/h2*1H;1-2H2

InChI key

LIAWOTKNAVAKCX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Hydrazine dihydrochloride can be used as a reducing agent in the conversion of carbonyls to methylene compounds and α, β-epoxyketones to allylic alcohols. It can also reduce alkenes, alkynes, and nitro groups. In addition, it is used to synthesize hydrazides and dinitrogen-containing heterocycles.

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Hydrazine
Roden B, et al.
e-EROS Encyclopedia of Reagents for Organic Synthesis, 1-7 (2001)
Rapid synthesis of 2, 5-disubstituted 1, 3, 4-oxadiazoles under microwave irradiation.
Bentiss F, et al.
Synthetic Communications, 31(6), 935-938 (2001)
Spectrophotometric method for determination of hydrazine.
Watt GW and Chrisp JD.
Analytical Chemistry, 24(12), 2006-2008 (1952)
Accelerated synthesis of 3, 5-disubstituted 4-amino-1, 2, 4-triazoles under microwave irradiation.
Bentiss F, et al.
Tetrahedron Letters, 41(10), 1539-1541 (2000)
A silver colloid produced by reduction with hydrazine as support for highly sensitive surface-enhanced Raman spectroscopy
Nickel U, et al.
Langmuir, 16(23), 9087-9091 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service