Skip to Content
Merck
All Photos(3)

Documents

185698

Sigma-Aldrich

Barbituric acid

ReagentPlus®, 99%

Synonym(s):

2,4,6-Trihydroxypyrimidine, Malonylurea

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H4N2O3
CAS Number:
Molecular Weight:
128.09
Beilstein:
120502
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

Quality Level

product line

ReagentPlus®

Assay

99%

form

powder

mp

248-252 °C (dec.) (lit.)

SMILES string

O=C1CC(=O)NC(=O)N1

InChI

1S/C4H4N2O3/c7-2-1-3(8)6-4(9)5-2/h1H2,(H2,5,6,7,8,9)

InChI key

HNYOPLTXPVRDBG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Barbituric acid is a useful acid for organic and drug syntheses. Its dihydrate form can be synthesized from barbituric acid via crystallization from aqueous solution. Crystal structure of barbituric acid (in tautomeric form) has been investigated by a three dimensional fourier transform method. Its enol crystal form has been reported to be thermodynamically stable.

Application

Barbituric acid (BA) may be used in the preparation of the corresponding hemiaminals, via chemoselective reduction in the presence of SmI2/H2O reagent. It may be used in the preparation of BA- modified conjugated carbon nitride nanosheets.
It may be used to synthesize:
  • 5-ylidenebarbituric acid derivatives via Knoevenagel condensation with aromatic and α,β-conjugated aromatic aldehydes
  • 5-diaminomethylenebarbiturates by reacting with substituted carbodiimides

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

302.0 °F - closed cup

Flash Point(C)

150.00 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Madalynn G Marshall et al.
Angewandte Chemie (International ed. in English), 55(4), 1309-1312 (2015-12-15)
The thermodynamically stable enol crystal form of barbituric acid, previously prepared as powder by grinding or slurry methods, has been obtained as single crystals by slow cooling from methanol solution. The selection of the enol crystal was facilitated by a
Selective reduction of barbituric acids using SmI2/H2O: synthesis, reactivity, and structural analysis of tetrahedral adducts.
Michal Szostak et al.
Angewandte Chemie (International ed. in English), 52(48), 12559-12563 (2013-10-15)
The crystal structure of anhydrous barbituric acid.
Bolton W.
Acta Crystallographica, 16(3), 166-173 (1963)
Preparation of 5-diaminomethylenebarbiturates by barbituric acid addition to carbodiimides.
Jursic BS et al.
Tetrahedron, 59(19), 3427-3432 (2003)
A simple method for knoevenagel condensation of a, ?-conjugated and aromatic aldehydes with barbituric acid.
Jursic BS.
Journal of Heterocyclic Chemistry, 38(3), 655-657 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service