Skip to Content
Merck
All Photos(3)

Documents

122777

Sigma-Aldrich

Alizarin

Dye content 97 %

Synonym(s):

1,2-Dihydroxyanthraquinone, Mordant Red 11

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H8O4
CAS Number:
Molecular Weight:
240.21
Colour Index Number:
58000
Beilstein:
1914037
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

form

solid

Quality Level

composition

Dye content, 97%

color

yellow

visual transition interval

10.1-12.1, red to violet(alkaline)
5.5-6.8, yellow to red(acid)

mp

279-283 °C (lit.)

solubility

0.1 M NaOH: 0.01 g/L

λmax

567 nm (2nd)
609 nm

ε (extinction coefficient)

13000 at 227-233 nm in 0.1 M NaOH at 0.01 g/L
26000 at 268-274 nm in 0.1 M NaOH at 0.01 g/L

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

Oc1ccc2C(=O)c3ccccc3C(=O)c2c1O

InChI

1S/C14H8O4/c15-10-6-5-9-11(14(10)18)13(17)8-4-2-1-3-7(8)12(9)16/h1-6,15,18H

InChI key

RGCKGOZRHPZPFP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Alizarin has been used as a biological stain to dye cotton fabrics and is used as standard in UV-visible spectrophotometer.

Biochem/physiol Actions

Alizarin (1, 2-dihydroxyanthraquinone) is an anthraquinone dye and acts as a main chromophore of madder dye. It is used as a dye in the production of lake pigments. Alizarin is a key constituent in extracts of roots of various members of Rubiaceae, Morinda, Gallium, Oldenlandia families.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 1 - Eye Dam. 1 - Skin Sens. 1A

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Nicholas Eastaugh, Valentine Walsh, Tracey Chaplin, Ruth Siddall
Pigment Compendium: A Dictionary and Optical Microscopy of Historical Pigments, By Nicholas Eastaugh, Valentine Walsh, Tracey Chaplin (2007)
Björn Hegner et al.
Scientific reports, 8(1), 12087-12087 (2018-08-16)
Severe vascular calcification develops almost invariably in chronic kidney patients posing a substantial risk to quality of life and survival. This unmet medical need demands identification of novel therapeutic modalities. We aimed to pinpoint components of the uremic microenvironment triggering
Tom Geerinckx et al.
Physiological and biochemical zoology : PBZ, 85(5), 481-490 (2012-08-21)
Teeth are generally used for actions in which they experience mainly compressive forces acting toward the base. The ordered tooth enamel(oid) and dentin structures contribute to the high compressive strength but also to the minor shear and tensile strengths. Some
Huiwu Long et al.
Advanced science (Weinheim, Baden-Wurttemberg, Germany), 5(3), 1700634-1700634 (2018-03-30)
Aqueous sodium-ion battery of low cost, inherent safety, and environmental benignity holds substantial promise for new-generation energy storage applications. However, the narrow potential window of water and the enlarged ionic radius because of hydration restrict the selection of electrode materials
SERS detection of red organic dyes inAg-agar gel.
Lofrumento C
Journal of Raman Spectroscopy, 44, 47-54 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service