Skip to Content
Merck
All Photos(3)

Key Documents

PP64

Sigma-Aldrich

IgG, Rabbit

Rabbit IgG Polyclonal Antibody validated for use in ELISA.

Synonym(s):

Rabbit Antibody, Rabbit IgG Antibody, Rabbit Immunoglobulin G

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

purified immunoglobulin

clone

polyclonal

manufacturer/tradename

Chemicon®

concentration

10 mg/mL

technique(s)

ELISA: suitable

shipped in

dry ice

target post-translational modification

unmodified

General description

Purified rabbit IgG.
Source: Normal rabbit serum collected from healthy normal rabbits.

Quality

Purity determined by gel electrophoresis to be greater than 95%.

Physical form

Format: Purified
Frozen nonsterile liquid in a 0.01 M Phosphate buffered saline, pH 7.1. Filtered through a 0.45 micron membrane filter.
IgG is purified by cohn fractionation followed by secondary purification with sodium caprylate.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 12 months. Avoid multiple freeze-thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Connective Tissue Growth Factor (CTGF/CCN2) enhances lactogenic differentiation of mammary epithelial cells via integrin-mediated cell adhesion.
Morrison, BL; Jose, CC; Cutler, ML
BMC Cell Biology null
Evidence that Igf2 down-regulation in postnatal tissues and up-regulation in malignancies is driven by transcription factor E2f3.
Lui, JC; Baron, J
Proceedings of the National Academy of Sciences of the USA null
Functional dissection of the NuA4 histone acetyltransferase reveals its role as a genetic hub and that Eaf1 is essential for complex integrity.
Mitchell, L; Lambert, JP; Gerdes, M; Al-Madhoun, AS; Skerjanc, IS; Figeys, D; Baetz, K
Molecular and cellular biology null
Polarized release of human cytomegalovirus from placental trophoblasts.
Hemmings, DG; Guilbert, LJ
Journal of virology null
Chromatin immunoprecipitation (ChIP): revisiting the efficacy of sample preparation, sonication, quantification of sheared DNA, and analysis via PCR.
Schoppee Bortz, PD; Wamhoff, BR
Testing null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service