Skip to Content
Merck
All Photos(1)

Key Documents

OP188

Sigma-Aldrich

Anti-FAP Mouse mAb (F11-24)

liquid, clone F11-24, Calbiochem®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

F11-24, monoclonal

form

liquid

does not contain

preservative

species reactivity

human

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG1

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FAP(2191)

General description

Anti-FAP, mouse monoclonal, clone F11-24, recognizes fibroblast activation protein in human. It is validated for use in flow cytometry, immunoprecipitation, & immunohistochemistry (frozen sections).
Protein A purified mouse monoclonal antibody. Recognizes human fibroblast activation protein (FAP).
Recognizes fibroblast activation protein (FAP).

Immunogen

HEK293 cells transfected with an expression vector encoding full-length human FAP
Human

Application

Flow Cytometry (see comments)

Frozen Sections (1-5 µg/ml)

Immunoprecipitation (see comments)

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In PBS.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Analysis Note

Positive Control
Reactive human fibroblasts, glucagon-producing A cells

Other Notes

Does not react with resting fibroblasts of normal organ mesenchyme or parenchymal cells, except for the glucagon-producing A cells found in the pancreatic islets. This antibody has also been reported to work for flow cytometry and immunoprecipitation. Antibody should be titrated for optimal results in individual systems.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Larissa Belov et al.
Methods in molecular biology (Clifton, N.J.), 1619, 263-301 (2017-07-05)
DotScan antibody microarrays were initially developed for the extensive surface profiling of live leukemia and lymphoma cells. DotScan's diagnostic capability was validated with an extensive clinical trial using mononuclear cells from the blood or bone marrow of leukemia or lymphoma
Marta Trüb et al.
Journal for immunotherapy of cancer, 8(2) (2020-07-04)
The costimulatory receptor 4-1BB (CD137, TNFRSF9) plays an important role in sustaining effective T cell immune responses and is investigated as target for cancer therapy. Systemic 4-1BB directed therapies elicit toxicity or low efficacy, which significantly hampered advancement of 4-1BB-based
Sarah M Ridge et al.
International journal of cancer, 142(10), 2056-2067 (2017-12-22)
Mesenchymal stem cells (MSCs) are a heterogeneous population of multipotent cells that are capable of differentiating into osteocytes, chondrocytes and adipocytes. Recently, MSCs have been found to home to the tumour site and engraft in the tumour stroma. However, it

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service