Skip to Content
Merck
All Photos(2)

Documents

HMMP1MAG-55K

Millipore

MILLIPLEX® Human MMP Magnetic Bead Panel 1 - Immunology Multiplex Assay

Matrix Metalloproteinase Bead-Based Multiplex Assays using the Luminex technology enable the simultaneous analysis of multiple MMPs biomarkers in human serum, plasma and cell culture samples.

Synonym(s):

Human matrix metalloproteinase multiplex kit, Luminex® Human MMP immunoassay, Millipore human MMP panel

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.84

Quality Level

species reactivity

human

manufacturer/tradename

Milliplex®

assay range

standard curve range: 146-150,000 pg/mL
(MMP-3)

standard curve range: 58-60,000 pg/mL
(MMP-13)

standard curve range: 98-100,000 pg/mL
(MMP-12)

technique(s)

multiplexing: suitable

detection method

fluorometric (Luminex xMAP)

shipped in

wet ice

General description

MMPs (matrix metalloproteinases), a family of zinc proteases responsible for the breakdown of extracellular matrix (ECM), play a key role in normal physiological processes, such as embryonic development and tissue morphogenesis, tissue and bone remodeling, wound healing, and angiogenesis. These processes rely on MMPs′ role in the cleavage of cell surface receptors, the release of apoptotic ligands, cell proliferation and differentiation, and chemokine activity modulation. Similar in structure, MMPs are synthesized and secreted as inactive pro-enzymes that require proteolytic cleavage for activation. This process can be mediated by serine proteases or other MMPs. An increase in MMP expression occurs in response to a wide range of stimuli, including adhesion molecules, growth factors, cytokines, and hormones. Regulation of MMP activity is controlled primarily by TIMPs (tissue inhibitors of metalloproteinases). Therefore, disruption of the MMP/TIMP balance can result in arthritis, cardiovascular disease and tumor growth and metastasis.

MILLIPLEX® Human MMP Bead Panel 1 is a 3 plex kit to be used for the simultaneous quantification of any or all the following analytes in serum/plasma samples: MMP-3, MMP-12, and MMP-13.

The Luminex® xMAP® platform uses a magnetic bead immunoassay format for ideal speed and sensitivity to quantitate multiple analytes simultaneously, dramatically improving productivity while conserving valuable sample volume.

Panel Type: Cytokines/Chemokines

Specificity

Cross-reactivity between the antibodies and any of the other analytes in this panel is non-detectable or negligible.

Application

  • Analytes: MMP-3, MMP-12, MMP-13
  • Recommended Sample type: Serum and plasma; this panel has also been analytically verified in urine, tissue/cell lystate and culture supernatant samples
  • Recommended Sample dilution: Neat
  • Assay Run Time: One day
  • Research Category: Inflammation & Immunology

Features and Benefits

Design your multiplex kit by choosing available analytes within this panel.

Storage and Stability

Recommended storage for kit components is 2 - 8°C.

Other Notes

Sensitivity: See protocol for sensitivities of individual analytes

Legal Information

Luminex is a registered trademark of Luminex Corp
MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
xMAP is a registered trademark of Luminex Corp

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Sens. 1 - STOT RE 2

Target Organs

Respiratory Tract

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Saritha Tantravedi et al.
Oncotarget, 8(70), 115280-115289 (2018-02-01)
When crypt stem cells of the gastrointestinal tract become injured, the result is increased synthesis of pro-inflammatory cytokines and matrix metalloproteinases by their progeny - the colonic epithelium. Chronic inflammation of the gastrointestinal tract is a characteristic of inflammatory bowel
Karolina Minta et al.
Scientific reports, 10(1), 18075-18075 (2020-10-24)
Matrix metalloproteinases (MMPs) are extracellular enzymes involved in the degradation of extracellular matrix (ECM) proteins. Increased expression of MMPs have been described in traumatic brain injury (TBI) and may contribute to additional tissue injury and blood-brain barrier damage. The objectives
Karolina Minta et al.
Clinica chimica acta; international journal of clinical chemistry, 512, 74-83 (2020-12-05)
Altered levels of two extracellular matrix (ECM) proteoglycans, brevican and neurocan, have been found in brain injury models; however, their proteolytic processing in traumatic brain injury (TBI) remains unexplored. A disintegrin and metalloproteinase with thrombospondin motifs (ADAMTS) is a possible
Yingqian Zhang et al.
Journal of cerebral blood flow and metabolism : official journal of the International Society of Cerebral Blood Flow and Metabolism, 37(8), 2963-2974 (2016-11-26)
Blood-brain barrier (BBB) disruption plays an important role in pathophysiological progress of ischemic stroke. However, our knowledge of the dynamic change of BBB permeability and its mechanism remains limited. In the current study, we used a non-human primate (NHP) MCAO
Jonathan H Soslow et al.
Journal of cardiac failure, 25(4), 259-267 (2019-02-15)
Cardiomyopathy is the leading cause of death in Duchenne muscular dystrophy (DMD). Standard cardiac biomarkers are poor indicators of DMD cardiovascular disease. Matrix metalloproteinases (MMPs) and tissue inhibitors of metalloproteinases (TIMPs) regulate collagen turnover. Given the cardiac fibrosis seen in

Related Content

Simplify protein biomarker discovery with the MILLIPLEX® PLEXpedition 115-Plex Multiplex Screening Panel. Effortlessly screen cytokines and other markers with Luminex® technology.

Simplify protein biomarker discovery with the MILLIPLEX® PLEXpedition 115-Plex Multiplex Screening Panel. Effortlessly screen cytokines and other markers with Luminex® technology.

Simplify protein biomarker discovery with the MILLIPLEX® PLEXpedition 115-Plex Multiplex Screening Panel. Effortlessly screen cytokines and other markers with Luminex® technology.

Simplify protein biomarker discovery with the MILLIPLEX® PLEXpedition 115-Plex Multiplex Screening Panel. Effortlessly screen cytokines and other markers with Luminex® technology.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service