Skip to Content
Merck
All Photos(1)

Documents

AP200P

Sigma-Aldrich

Goat Anti-Mouse light chain Antibody, HRP conjugate

Chemicon®, from goat

Synonym(s):

Goat Anti-Mouse HRP, HRP Goat Anti-Mouse, Mouse Light Chain Detection

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

goat

Quality Level

conjugate

peroxidase conjugate

antibody form

affinity purified immunoglobulin

antibody product type

secondary antibodies

clone

polyclonal

species reactivity

mouse

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
western blot: suitable

isotype

IgG

shipped in

wet ice

target post-translational modification

unmodified

General description

Antibody molecules typically comprise two immunoglobulin light chains covalently bound to a pair of heavy chains. Immunoglobulin light chains occur in two types, designated by the Greek letters kappa and lambda. Kappa and gamma light chains are approximately 250 amino acids in length with an average mass of about 25 kDa. The ratio of kappa to lambda found in the immunoglobulin population varies by species.

Specificity

Minimal cross-reaction with bovine, goat, horse, human, rabbit, rat, and sheep.

Application

Goat anti-Mouse light chain Antibody, HRP conjugate detects level of Mouse light chain & has been published & validated for use in ELISA & WB.

Target description

25 kDa

Physical form

Lyophilized powder from a solution with 0.01M Sodium Phosphate, 0.25M NaCl, pH 7.6. 15 mg/mL BSA as stabilizer. Purified by immunoaffinity chromatography.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Koki Ikemoto et al.
Acta histochemica et cytochemica, 54(2), 65-72 (2021-05-21)
Spontaneous Raman spectroscopy, which senses changes in cellular contents of reduced cytochrome c, could be a powerful tool for label-free evaluation of ischemic hearts. However, undetermined is whether it is applicable to evaluation of myocardial viability in ischemic hearts. To
Edward N Anderson et al.
The Journal of biological chemistry, 292(47), 19160-19178 (2017-09-20)
The family of TGF-β and bone morphogenetic protein (BMP) signaling proteins has numerous developmental and physiological roles. They are made as proprotein dimers and then cleaved by proprotein convertases to release the C-terminal domain as an active ligand dimer. Multiple
Daiki Nakatsu et al.
Genes to cells : devoted to molecular & cellular mechanisms, 19(7), 565-581 (2014-06-04)
Tricellular tight junctions (tTJs) are specialized structural variants of tight junctions within tricellular contacts of an epithelial sheet and comprise several transmembrane proteins including lipolysis-stimulated lipoprotein receptor (angulin-1/LSR) and tricellulin. To elucidate the mechanism of its formation, we carried out
Hristo Georgiev et al.
Frontiers in immunology, 9, 714-714 (2018-04-25)
Follicular helper (TFH) and regulatory (TFR) cells are critical players in managing germinal center (GC) reactions that accomplish effective humoral immune responses. Transcriptome analyses were done comparing gene regulation of TFH and TFR cells isolated from Peyer's Patches (PP) and
Angelica P Lopez et al.
Journal of molecular biology, 428(14), 2814-2831 (2016-05-25)
The lens epithelium-derived growth factor p75 (LEDGF/p75) is a chromatin-bound protein essential for efficient lentiviral integration. Genome-wide studies have located LEDGF/p75 inside actively transcribed genes where it mediates lentiviral integration. Although its role in HIV-1 integration is clearly established, the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service