Skip to Content
MilliporeSigma

ABS85

Anti-CPT2 Antibody

serum, from rabbit

Synonym(s):

Carnitine O-palmitoyltransferase 2, mitochondrial, Carnitine palmitoyltransferase II, CPT II

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
WB
Citations:
6
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat

species reactivity (predicted by homology)

human (immunogen homology)

technique(s)

western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CPT2(1376)

General description

Carnitine palmitoyltransferase II (CPT2, or CPT II) belongs to the carnitine/choline acetyltransferase family. It is a ubiquitous protein found in the inner membrane of the mitochondria and plays an essential role in fatty acid β-oxidation. CPT2 is part of the system involving CPT1 and carnitine/acylcarnitine translocase (CACT), that facilitate the transport of long chain fatty acids into the mitochondrial matrix. CPT2 plays the role of restoring these molecules to original esterified forms suitable for β-oxidation, after they arrive in the matrix. Defects in CPT2 has varying outcomes: in adults, it results in episodes of rhabdomyolysis (breakdown of muscle fibers which circulate in the bloodstream), after prolonged and vigorous exercise; and in infants, CPT2 deficiency induces hypoketotic hypoglycemia and eventual cardiac damage.
~68 kDa observed (cleaved form). Uniprot gives a calculated weight of 74 kDa; however, CPT2 has been observed at ~68 kDa (Faye, A, et al. (2007). The Journal of Biological Chemistry. 282(37):26908–26916.). Uncharacterized bands appear at ~55, 58, 72, and 100 kDa in some lysates.

Immunogen

KLH-conjugated linear peptide corresponding to human CPT2.

Application

Anti-CPT2 detects levels of CPT2 proteins & has been published & validated for use in WB.

Analysis Note

Control
Rat liver mitochondria tissue lysate
Evaluated by Western Blot in rat liver mitochondria tissue lysate.

Western Blot Analysis: A 1:1,000 dilution of this antibody detected CPT2 on 10 µg of rat liver mitochondria tissue lysate.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 1



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library