Skip to Content
Merck
All Photos(1)

Key Documents

AB3538P

Sigma-Aldrich

Anti-Monocarboxylate Transporter 1 Antibody

Chemicon®, from rabbit

Synonym(s):

MCT1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... SLC16A1(6566)

Specificity

Recognizes human Monocarboxylate Transporter 1 (MCT1). The immunogen shows no significant sequence homology with MCT2-8.

Immunogen

A 15 amino acid peptide sequence near the C-terminus of human MCT1 (Gerhart et al. 1997; Knott et al. 1999; Takanaga et al. 1995; Carpenter et al. 1996; Koehler-Stec et al. 1998; Yoon et al. 1999; Halestrap & Price 1999).

Application

Research Category
Neuroscience
Research Sub Category
Ion Channels & Transporters
This Anti-Monocarboxylate Transporter 1 Antibody is validated for use in ELISA, WB for the detection of Monocarboxylate Transporter 1.
Western blot: 1-5 μg/mL using ECL.
ELISA: 1:10,000-1:100,000 using 50-100 ng control peptide per well.
Optimal working dilutions must be determined by the end user.

Physical form

Affinity purified immunoglobulin. Liquid in PBS containing 0.1% BSA.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 6 months from date of receipt. Avoid repeated freeze/thaw cycles.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Expression of monocarboxylate transporters 1, 2, and 4 in human tumours and their association with CD147 and CD44.
Pinheiro, C; Reis, RM; Ricardo, S; Longatto-Filho, A; Schmitt, F; Baltazar, F
Journal of Biomedicine and Biotechnology null
Julieta Afonso et al.
Cell cycle (Georgetown, Tex.), 15(3), 368-380 (2015-12-05)
Monocarboxylate transporters (MCTs) are vital for intracellular pH homeostasis by extruding lactate from highly glycolytic cells. These molecules are key players of the metabolic reprogramming of cancer cells, and evidence indicates a potential contribution in urothelial bladder cancer (UBC) aggressiveness
S-Nitrosation of monocarboxylate transporter 1: inhibition of pyruvate-fueled respiration and proliferation of breast cancer cells.
Diers, AR; Broniowska, KA; Chang, CF; Hill, RB; Hogg, N
Free radical biology & medicine null
Chun-Kai Huang et al.
Nature communications, 8, 14706-14706 (2017-03-11)
Adipocytes are the most abundant stromal partners in breast tissue. However, the crosstalk between breast cancer cells and adipocytes has been given less attention compared to cancer-associated fibroblasts. Here we find, through systematic screening, that primary mammary gland-derived adipocytes (MGDAs)
Eduardo C A Silva et al.
Oncotarget, 9(29), 20386-20398 (2018-05-15)
Metabolic reprogramming is one of the hallmarks of cancer. The hyperglycolytic phenotype is often associated with the overexpression of metabolism-associated proteins, such as monocarboxylate transporters (MCTs). MCTs are little explored in germ cell tumors (GCTs), thus, the opportunity to understand

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service