Skip to Content
Merck
All Photos(1)

Key Documents

8.20315

Sigma-Aldrich

1-Chloronaphthalene

for synthesis

Synonym(s):

1-Chloronaphthalene, α-Naphthyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H7Cl
CAS Number:
Molecular Weight:
162.62
MDL number:
UNSPSC Code:
12352101
EC Index Number:
201-967-3
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

0.05 hPa ( 20 °C)

Quality Level

Assay

85.0-88.0% (GC)

form

liquid

autoignition temp.

>500 °C

potency

1540 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rat)

refractive index

n20/589 1.6332

pH

7 (20 °C in H2O, undiluted)

bp

259 °C/1013 hPa

mp

-6--2.5 °C

transition temp

flash point 115 °C

density

1.19 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

Clc1c2c(ccc1)cccc2

InChI

1S/C10H7Cl/c11-10-7-3-5-8-4-1-2-6-9(8)10/h1-7H

InChI key

JTPNRXUCIXHOKM-UHFFFAOYSA-N

Application

  • Enhancement of polymer solar cells: 1-Chloronaphthalene was used as a solid additive in high-efficiency polymer solar cells developed with a benzo-difuran-based donor polymer, improving device performance and stability (Chen et al., 2023).
  • Study of charge transfer complexes: The electronic and vibrational properties of tetracyanoethylene-chloronaphthalene charge transfer complex in solution were explored using TD-DFT and ab initio molecular dynamics, providing insights into molecular interactions and stability (Coppola et al., 2022).
  • Efficiency boost in solar cells: 1-Chloronaphthalene was employed as a nontoxic, halogen-free additive in polymer solar cells, significantly boosting their efficiency and highlighting its potential as an environmentally friendly material for solar energy applications (Liu et al., 2021).
  • Development of nonhalogenated-solvent solar cells: Medium-band-gap small-molecule acceptors based on a 6,12-dihydro-diindolo[1,2-b:1,2-e]pyrazine unit were used in nonhalogenated-solvent-processed polymer solar cells, where 1-Chloronaphthalene acted as a solvent, promoting higher device efficiencies and reduced environmental impact (Chen et al., 2019).

Analysis Note

Assay (GC, area%): 85.0 - 88.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.188 - 1.195
2-Chloronaphthalene (GC, area%): 12.0 - 15.0 % (a/a)
Identity (IR): passes test

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

249.8 °F - closed cup

Flash Point(C)

121 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service