Skip to Content
Merck
All Photos(1)

Key Documents

506119

Sigma-Aldrich

PhosphoDetect Anti-p38 MAP Kinase (pThr¹⁸⁰, pTyr¹⁸²) Rabbit pAb

liquid, Calbiochem®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

liquid

contains

≤0.1% sodium azide as preservative

species reactivity

rat, mouse, canine, human

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pThr180/pTyr182)

Gene Information

human ... MAPK14(1432)

General description

Note: 1 T = 1 test. Sufficient for 10 Western miniblots.
Rabbit polyclonal antibody adsorbed against non-phosphopeptide corresponding to the immunogen phosphorylation site, followed by immunoaffinity chromatography. Recognizes the ~p38 MAPK phosphorylated at Thr180 and Tyr182.
Recognizes p38α, β, and γ/ERK6 phosphorylated at Thr180 and Tyr182 in uv-treated HEK293 cells and sorbitol-treated PC12 cells.
This PhosphoDetect Anti-p38 MAP Kinase Rabbit pAb is validated for use in WB, ICC for the detection of p38 MAP Kinase .

Immunogen

Human
a synthetic phosphopeptide corresponding to amino acids surrounding the Thr¹⁸⁰ and Tyr¹⁸² phosphorylation sites of human p38 MAP Kinase

Application

Immunoblotting (1:1000)

Immunocytochemistry (see comments)

Warning

Toxicity: Standard Handling (A)

Physical form

In Dulbecco′s PBS (without Mg2+ and Ca2+), 1 mg/ml BSA, 50% glycerol, pH 7.3.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Analysis Note

Positive Control
UV irradiated HEK293 cells or sorbitol-treated PC12 cells

Other Notes

Does not appreciably recognize the corresponding phosphorylated forms of either p42/44 MAPK or SAPK/JNK MAPK. The immunogen sequence is 100% conserved in p38α, β, and γ. The immunogen sequence is also 100% conserved in mouse, moneky, and carp p38, but cross-reactivity has not been tested. This antibody has also been reported to work for immunocytochemistry. When diluted 1:1000, there is sufficient antibody for 10 immunoblots at 10 ml per blot. Variables associated with assay conditions will dictate the proper working dilution.
Marinissen, M.J., et al. 2001. Genes Dev.15, 535.
Nakagami, H., et al. 2001. Diabetes50, 1472.
Wilsbacher, J.L., et al. 1999. J. Biol. Chem.274, 16988.
Raingeaud, J., et al. 1995. J. Biol. Chem.270, 7420.
Zervos, A.S., et al. 1995. Proc. Natl. Acad. Sci. USA92, 10531.
Han, J., et al. 1994. Science265, 808.
Lee, J.C., et al. 1994. Nature372, 739.
Rouse, J., et al. 1994. Cell78, 1027.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Kaufmann et al.
Acta physiologica (Oxford, England), 213(4), 920-932 (2015-01-17)
Hypoxia and sympathetic activation are main factors in the pathogenesis of acute kidney injury (AKI). We tested the hypothesis that noradrenaline (NE) in combination with hypoxia aggravates the vasoreactivity of renal arteries after hypoxia/re-oxygenation (H/R). We tested the role of
Noriyuki Shibata et al.
Neuropathology : official journal of the Japanese Society of Neuropathology, 28(4), 387-398 (2008-03-04)
Emerging evidence suggests the involvement of programmed cell death and inflammation in amyotrophic lateral sclerosis (ALS). To assess molecular pathological effects of the anti-inflammatory peroxisome proliferator-activated receptor-gamma (PPARgamma) agonist pioglitazone in ALS, we verified changes in the population of neurons
Andreas Patzak et al.
American journal of physiology. Regulatory, integrative and comparative physiology, 293(6), R2232-R2242 (2007-09-28)
Adenosine (Ado) enhances ANG II-induced constrictions of afferent arterioles (Af) by receptor-dependent and -independent pathways. Here, we test the hypothesis that transient Ado treatment has a sustained effect on Af contractility, resulting in increased ANG II responses after longer absence
Douglas W Strand et al.
American journal of clinical and experimental urology, 2(3), 239-248 (2014-11-07)
Transforming Growth Factor-β (TGF-β) regulates the reactive stroma microenvironment associated with most carcinomas and mediates expression of many stromal derived factors important for tumor progression, including FGF-2 and CTGF. TGF-β is over-expressed in most carcinomas, and FGF-2 action is important
En Yin Lai et al.
Kidney international, 76(9), 953-959 (2009-07-25)
Many agents constrict isolated afferent arterioles only at concentrations higher than their physiological levels. Here we determined if norepinephrine, as released by sympathetic nerve activity, could influence the angiotensin II responsiveness of isolated mouse afferent arterioles. Pretreatment of the arterioles

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service