Skip to Content
Merck
All Photos(1)

Key Documents

219373

Sigma-Aldrich

Cathepsin G, Human Neutrophil

Cathepsin G, Human Neutrophil, CAS 107200-92-0, is a purified native cathepsin G. Acts as a potent agonist of human platelet activation leading to their aggregation.

Synonym(s):

Cathepsin G, Human Neutrophil

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77

biological source

human neutrophils

Quality Level

Assay

≥95% (SDS-PAGE)

form

lyophilized solid (Salt-free)

specific activity

≥2 units/mg protein

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

technique(s)

inhibition assay: suitable

suitability

suitable for molecular biology

application(s)

life science and biopharma

shipped in

ambient

storage temp.

−20°C

Gene Information

human ... CTSG(1511)

General description

Research area: Cell Signaling

Cathepsin G is stored in its active form in azurophil granules of neutrophils or monocytes. Its mature form contains one potential glycan-binding site and three disulfide bonds.

Application

Cathepsin G, Human Neutrophil has been used to determine the in vitro bactericidal activities of cathepsin G and for in vitro digestion of plasma/acute wound fluids.

Biochem/physiol Actions

This protease cleaves the triple-helix structure of type I collagen in a restricted manner and degrades nonhelical regions of type I and II collagens, fibronectin, aggrecan, and elastin. Additionally, cathepsin G activates ProMMP-3 and -8. Cathepsin G serves as a chemoattractant for human phagocytes and enhances the random motility of T cells. Additionally, it initiates chemotaxis, activates ERK1/2 and p38 MAPK, and facilitates the translocation of PKCζ to the cell membrane, without inducing a detectable calcium flux. Other biological functions of cathepsin G include: ECM components and plasma proteins degradation, bactericidal actions, cleavage of inflammatory mediators, conversion of angiotensin I to angiotensin II, platelet activation and induction of airway submucosal gland secretion.

Warning

Toxicity: Standard Handling (A)

Unit Definition

One unit is defined as the amount of enzyme that will hydrolyze 1.0 µmol of Suc-AAPF-pNA per min at 25°C, pH 7.5.

Preparation Note

Prepared from blood that has been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV.

Reconstitute in 150 mM NaCl, 50 mM sodium acetate buffer, pH 5.5.

Reconstitution

Following reconstitution, aliquot and freeze (-70°C) for long term storage or refrigerate (4°C) for short term storage. Stock solutions are stable for up to 1 week at 4°C or for up to 6 months at -70°C.
Reconstitute in 50 mM NaOAc and 150 mM NaCl, pH 5.5.

Other Notes

Glusa, E., and Adam C. 2001. Por. J. Pharmacol.133, 422.
Shamamian, P., et al. 2001. J. Cell Physiol.189, 197.
Groutas, W.C., et al. 1993. Biochem. Biophys. Res. Commun.197, 730.
Stone, P.J., et al. 1993. Biochem. Biophys. Res. Commun.197, 130.
Groutas, W.C., et al. 1992. Arch. Biochem. Biophys.294, 144.
Maison, C.M., et al. 1991. J. Immunol.147, 921.
Travis, J. 1988. Am. J. Med.84, 37.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Isabelle Migeotte et al.
Cytokine & growth factor reviews, 17(6), 501-519 (2006-11-07)
The formyl peptide receptor (FPR) family is involved in host defence against pathogens, but also in sensing internal molecules that may constitute signals of cellular dysfunction. It includes three subtypes in human and other primates. FPR responds to formyl peptides
Dental and oral biology, biochemistry
Yamakoshi, Y, et al.
reference module in biomedical sciences (2014)
Brice Korkmaz et al.
Biochimie, 90(2), 227-242 (2007-11-21)
Polymorphonuclear neutrophils form a primary line of defense against bacterial infections using complementary oxidative and non-oxidative pathways to destroy phagocytized pathogens. The three serine proteases elastase, proteinase 3 and cathepsin G, are major components of the neutrophil primary granules that
Jun Cai et al.
iScience, 27(2), 109005-109005 (2024-02-09)
Endogenous and bacterial proteases play important roles in wound healing and infection. Analysis of alterations in the low-molecular-weight peptidome by individual enzymes could therefore provide insight into proteolytic events occurring in wounds and may aid in the discovery of biomarkers.
Dieke van der Windt et al.
FEMS immunology and medical microbiology, 66(3), 445-448 (2012-09-05)
Although the Streptococcus pneumoniae polysaccharide capsule is an important virulence factor, ~ 15% of carriage isolates are nonencapsulated. Nonencapsulated S. pneumoniae are a cause of mucosal infections. Recent studies have shown that neutrophils kill S. pneumoniae predominately through neutrophil proteases, such as elastase

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service