Skip to Content
Merck
All Photos(1)

Key Documents

20774

Millipore

IHC Select Streptavidin-HRP, prediluted

50 mL IHC Select Streptavidin-HRP, prediluted for IHC(P).

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116010
eCl@ss:
42029053

form

liquid

packaging

pkg of 50 mL

manufacturer/tradename

Chemicon®
IHC Select

technique(s)

immunohistochemistry: suitable (paraffin)

detection method

colorimetric

shipped in

wet ice

Application

Prediluted for use in Immunohistochemical staining procedures. For example:

Blocking Reagent

Primary Antibody or Negative Control Reagent

Biotinylated Secondary Antibody

Streptavidin-HRP Conjugate

Chromogen

Counterstain

For best results use this conjugate in conjunction with other IHC Select detection reagents. A complete list of reagents is available at www.chemicon.com.

Physical form

Dropper vial containing Streptavidin conjugated to Horseradish Peroxidase in a ready-to-use format

Storage and Stability

When stored at 2-8°C, this reagent is stable up to the expiration date printed on the label. Do not freeze or expose to elevated temperatures.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Erika Terzuoli et al.
Cancers, 11(12) (2019-12-11)
Melanoma and non-small-cell lung carcinoma (NSCLC) cell lines are characterized by an intrinsic population of cancer stem-like cells (CSC), and high expression of detoxifying isozymes, the aldehyde dehydrogenases (ALDHs), regulating the redox state. In this study, using melanoma and NSCLC
Ilianna Zoi et al.
Breast cancer research : BCR, 21(1), 132-132 (2019-12-05)
ERBB-2 is overexpressed in about 20% of breast cancers (BCs), indicating poor prognosis. The receptor activator of nuclear factor-κB (RANK) pathway is implicated in ERBB-2 (+) BC. The purpose of this study was to elucidate the underlying molecular mechanism of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service