Skip to Content
MilliporeSigma

Skip To

20-153

Chromatin Immunoprecipitation (ChIP) Dilution Buffer

For use in the Chromatin Immunoprecipitation Assays.

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
24 mL
Check Cart for Availability
$208.00

About This Item

UNSPSC Code:
41116012
NACRES:
NA.41
eCl@ss:
32160405

$208.00


Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

liquid

manufacturer/tradename

Upstate®

shipped in

wet ice

Application

Research Category
All

Physical form

0.01% SDS, 1.1% Triton X- 100, 1.2mM EDTA, 16.7mM Tris-HCl, pH 8.1, 167mM NaCl.

Preparation Note

1 year at 4°C

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
17-61117-1046017-610
manufacturer/tradename

Upstate®

manufacturer/tradename

-

manufacturer/tradename

-

manufacturer/tradename

Magna ChIP®

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

shipped in

wet ice

shipped in

-

shipped in

-

shipped in

-

form

liquid

form

-

form

-

form

-


pictograms

Exclamation mark

Storage Class

12 - Non Combustible Liquids

flash_point_f

Not applicable

flash_point_c

Not applicable

signalword

Danger

Hazard Classifications

Aquatic Chronic 3 - ED ENV 1 - Eye Irrit. 2



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters