Skip to Content
Merck
All Photos(1)

Key Documents

12-515

Sigma-Aldrich

Normal Sheep IgG

Normal Sheep IgG Polyclonal Antibody control validated for use in Western Blotting.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

sheep

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

clone

polyclonal

manufacturer/tradename

Upstate®

technique(s)

western blot: suitable

shipped in

dry ice

target post-translational modification

unmodified

Application

Research Category
Secondary & Control Antibodies
Research Sub Category
Whole Immunoglobulin Secondary Antibodies

Quality

Routinely evaluated as a negative non-specific IgG control in assays that require sheep derived antibodies.

Physical form

0.02M potassium phosphate, 0.15M NaCl, pH 7.2, with 0.01% sodium azide

Storage and Stability

2 years at -20°C

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sheila A Sweeney et al.
Biology of reproduction, 72(1), 172-178 (2004-09-24)
Betaglycan was originally characterized as the type III receptor for TGFbeta, yet recent research has indicated that betaglycan can serve as an accessory receptor for inhibin. To understand better the action of inhibin in avian follicular development, we have investigated
Dominique Mayrand et al.
Methods in molecular biology (Clifton, N.J.), 1773, 93-105 (2018-04-25)
Optimal imaging methods are necessary in order to perform a detailed characterization of thick tissue samples from either native or engineered tissues. Tissue-engineered substitutes are featuring increasing complexity including multiple cell types and capillary-like networks. Therefore, technical approaches allowing the
Yunlong Yang et al.
Nature communications, 7, 11385-11385 (2016-05-07)
Signalling molecules and pathways that mediate crosstalk between various tumour cellular compartments in cancer metastasis remain largely unknown. We report a mechanism of the interaction between perivascular cells and tumour-associated macrophages (TAMs) in promoting metastasis through the IL-33-ST2-dependent pathway in
Liang Weng et al.
PLoS biology, 16(3), e2005090-e2005090 (2018-03-15)
Amino acid signaling mediated by the activation of mechanistic target of rapamycin complex 1 (mTORC1) is fundamental to cell growth and metabolism. However, how cells negatively regulate amino acid signaling remains largely unknown. Here, we show that interaction between 4F2
Amila Suraweera et al.
Communications biology, 4(1), 484-484 (2021-04-21)
Genomic stability is critical for normal cellular function and its deregulation is a universal hallmark of cancer. Here we outline a previously undescribed role of COMMD4 in maintaining genomic stability, by regulation of chromatin remodelling at sites of DNA double-strand

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service