1.16885
Fractogel® EMD TMAE Medcap (M)
Synonym(s):
Biochromatography, Fractogel® Ion exchangers, Ion exchanger for LC (Fractogel® TSK/EMD), Fractogel® EMD TMAE Medcap (M)
About This Item
Recommended Products
Quality Level
ligand
(Trimethylammoniumethyl)
description
strong anion exchanger, suspension in 20% ethanol and 150 mM NaCl (40-90 µm)
sterility
sterile (Caustic Stable)
product line
Fractogel®
form
resin
parameter
180 cm/hr flow rate
8 bar max. pressure
matrix active group
methacrylate
mean particle size
40-90 μm
capacity
150 mg binding capacity (BSA/mg of resin)
transition temp
flash point 35 °C (calculated)
density
1.430 g/cm3 at 20 °C
bulk density
1000 kg/m3
application(s)
plasma purification
recombinant protein
vaccine development
separation technique
anion exchange
strong anion exchange
storage temp.
2-30°C
Looking for similar products? Visit Product Comparison Guide
General description
Features and Benefits
- Excellent binding at high flow rates
- Excellent pressure-flow behavior
- Compatibility with 2.5 % (v/v) aqueous benzyl alcohol containing 150 mM NaCl storage solution
Packaging
- 1.16885.0100: Fractogel® EMD TMAE Medcap (M) Resin 100ml
- 1.16885.0010: Fractogel® EMD TMAE Medcap (M) Resin 10ml
- 1.16885.0500: Fractogel® EMD TMAE Medcap (M) Resin 500ml
- 1.16885.5000: Fractogel® EMD TMAE Medcap (M) Resin 5L
Legal Information
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Liq. 3
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(F)
95.0 °F
Flash Point(C)
35 °C
What is the Emprove® Program?
The Emprove® Program is a system providing comprehensive and thorough documentation of our filters and single-use components, pharma raw materials, and starting materials. Four document types are included with an Emprove® Program subscription:
Free of charge - Sign in to download
Free of charge - Sign in to download
Available by subscription or for a fee
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service