Skip to Content
Merck
All Photos(1)

Documents

1.12310

Supelco

Silicon standard solution

traceable to SRM from NIST acidic, (NH₄)₂SiF₆ in H₂O 1000 mg/l Si Certipur®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

Agency

suitable for EPA 200.7

product line

Certipur®

concentration

1000 mg/L Si in H2O

technique(s)

ICP: suitable

pH

3.4 (20 °C in H2O)

density

1.00 g/cm3 at 20 °C

application(s)

industrial qc
pharmaceutical

format

single component solution

storage temp.

15-25°C

Related Categories

Application


  • Particle size analysis for non-porous and mesoporous silica: Single-Particle Inductively Coupled Plasma Mass Spectrometry (SP-ICP-MS) is utilized to measure particle size and detect bound proteins on non-porous and mesoporous silica microspheres, highlighting its precision in assessing silicon-based carriers in biochemical applications (Miyashita et al., 2024).

  • Optimization of thermo-optic phase-shifters: The application of silicon standard solutions is crucial in optimizing double titanium nitride thermo-optic phase-shifter heaters using SOI technology, underscoring the importance of precise silicon calibration standards in semiconductor manufacturing (Krause and Malka, 2023).

Analysis Note

Concentration     β (Si): 990 - 1010 mg/l
Determination method: acidimetric titration.
(traceable to NIST - SRM 723e)
Accuracy of the method: ± 2 mg/l

Legal Information

CERTIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Rapid and sensitive test for silicate-induced scale deposits ensures industrial plant efficiency and sustainability.

Rapid and sensitive test for silicate-induced scale deposits ensures industrial plant efficiency and sustainability.

Rapid and sensitive test for silicate-induced scale deposits ensures industrial plant efficiency and sustainability.

Rapid and sensitive test for silicate-induced scale deposits ensures industrial plant efficiency and sustainability.

Related Content

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service