If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf
Select a Size
| Pack Size | SKU | Availability | Price |
|---|
About This Item
grade
ACS reagent
Quality Level
agency
reag. ISO, reag. Ph. Eur.
vapor density
4.1 (vs air)
vapor pressure
160 mmHg ( 20 °C)
product line
EMSURE®
assay
≥99.8%, 90-99.4% (GC)
form
liquid
potency
695 mg/kg LD50, oral (Rat), >3980 mg/kg LD50, skin (Rabbit)
contains
0.6-1.0% Ethanol as stabilizer
dilution
(for analytical testing)
refractive index
n20/D 1.445 (lit.)
bp
60.5-61.5 °C (lit.)
mp
−63 °C (lit.)
density
1.492 g/mL at 25 °C (lit.)
storage temp.
2-30°C
SMILES string
ClC(Cl)Cl
InChI
1S/CHCl3/c2-1(3)4/h1H
InChI key
HEDRZPFGACZZDS-UHFFFAOYSA-N
General description
Application
Other Notes
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| grade ACS reagent | grade - | grade - | grade - |
| assay ≥99.8%, 90-99.4% (GC) | assay ≥99.8% chloroform + ethanol basis | assay 99.0-99.4% (GC) | assay 99.0-99.4% (GC) |
| Quality Level 300 | Quality Level 200 | Quality Level - | Quality Level 200 |
| form liquid | form liquid | form liquid | form liquid |
| mp −63 °C (lit.) | mp −63 °C (lit.) | mp −63 °C (lit.) | mp −63 °C (lit.) |
| bp 60.5-61.5 °C (lit.) | bp 60.5-61.5 °C (lit.) | bp 60.5-61.5 °C (lit.) | bp 60.5-61.5 °C (lit.) |
Still not finding the right product?
Explore all of our products under Chloroform
signalword
Danger
Hazard Classifications
Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3
target_organs
Central nervous system, Liver,Kidney
Storage Class
6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects
wgk
WGK 3
flash_point_f
does not flash
flash_point_c
does not flash
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 1024454000 | 04022536893839 |
| 1024456025 | 04022536033648 |
| 1024451000 | 04022536033594 |
| 1024452500 | 04022536033617 |
| 1024456010 | 04022536033631 |
| 1024459190 | 04022536033662 |
-
How can I determine the shelf life / expiration / retest date of this product?
1 answer-
Helpful?
-
-
How is shipping temperature determined? And how is it related to the product storage temperature?
1 answer-
Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf
Helpful?
-


