Skip to Content
Merck
All Photos(1)

Key Documents

07-398

Sigma-Aldrich

Anti-phospho-NR2B (Ser1303) Antibody

Upstate®, from rabbit

Synonym(s):

Anti-DEE27, Anti-GluN2B, Anti-MRD6, Anti-NMDAR2B, Anti-NR2B, Anti-NR3, Anti-hNR3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat, human, mouse

manufacturer/tradename

Upstate®

technique(s)

western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

phosphorylation (pSer1303)

Gene Information

human ... GRIN2B(2904)
mouse ... Grin2B(14812)
rat ... Grin2B(24410)

Specificity

Recognizes phospho-NR2B.

Immunogen

Peptide corresponding to amino acids 1297-1309 of human phospho-NR2B

Application

Anti-phospho-NR2B (Ser1303) Antibody detects level of phospho-NR2B (Ser1303) & has been published & validated for use in WB.
Research Category
Neuroscience
Research Sub Category
Oxidative Stress

Neurodegenerative Diseases

Quality

routinely evaluated by immunoblot in a rat brain microsomal protein preparation

Target description

~180 kDa

Linkage

Replaces: 04-1148

Physical form

Format: Purified
Protein A Purified immunoglobulin in 30% glycerol, 0.07M Tris-glycine, pH 7.4, 0.105 M NaCl, 0.035% sodium azide as a preservative.
Protein A purified

Storage and Stability

Maintain for 2 years at -20°C from date of shipment. Aliquot to avoid repeated freezing and thawing. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Analysis Note

Control
Brain tissue

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Substrate-selective and calcium-independent activation of CaMKII by ?-actinin.
Jalan-Sakrikar, N; Bartlett, RK; Baucum, AJ; Colbran, RJ
The Journal of Biological Chemistry null
C Castillo et al.
Neuroscience, 177, 308-320 (2011-01-11)
The involvement of substance P (SP) in neuronal sensitization through the activation of the neurokinin-1-receptor (NK1r) in postsynaptic dorsal horn neurons has been well established. In contrast, the role of SP and NK1r in primary sensory dorsal root ganglion (DRG)
Roles of diverse glutamate receptors in brain functions elucidated by subunit-specific and region-specific gene targeting.
Mori, Hisashi and Mishina, Masayoshi
Life Sciences, 74, 329-336 (2003)
S Strack et al.
The Journal of biological chemistry, 275(31), 23798-23806 (2000-04-15)
Calcium influx through the N-methyl-d-aspartate (NMDA)-type glutamate receptor and activation of calcium/calmodulin-dependent kinase II (CaMKII) are critical events in certain forms of synaptic plasticity. We have previously shown that autophosphorylation of CaMKII induces high-affinity binding to the NR2B subunit of
Jamie McQueen et al.
eLife, 6 (2017-07-22)
Aberrant NMDA receptor (NMDAR) activity contributes to several neurological disorders, but direct antagonism is poorly tolerated therapeutically. The GluN2B cytoplasmic C-terminal domain (CTD) represents an alternative therapeutic target since it potentiates excitotoxic signaling. The key GluN2B CTD-centred event in excitotoxicity

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service