Skip to Content
Merck
All Photos(3)

Key Documents

06-720

Sigma-Aldrich

Anti-HDAC1 Antibody

Upstate®, from rabbit

Synonym(s):

histone deacetylase 1, reduced potassium dependency, yeast homolog-like 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat, human, mouse

packaging

antibody small pack of 25 μg

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable
immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... HDAC1(3065)
mouse ... Hdac1(433759)
rat ... Hdac1(297893)

General description

Mammalian HADC1 (also designated HD1), HDAC2 (also designated mammalian RPD3) and HDAC3, all are related to the yeast transcriptional regulator Rpd3p, are identified as histone deacetylases. The deacetylation of histones is associated with transcriptional silencing.

Specificity

Oher species cross-reactivity not tested.
Recognizes and is specific for HDAC1, Mr 65 kDa. May detects a non-specific protein at ~35 kDa.

Immunogen

6X-His-tagged fragment representing amino acids 53-482 of mouse histone deacetylase 1 (HDAC1).

Application

Immunoprecipitation:
5 μg of a previous lot immunoprecipitated HDAC1 from 500 μg of 3T3/A31 RIPA lysate.

Immunocytochemistry:
This antibody has been reported to immunostain HDAC1 in 1% paraformaldehyde fixed, 0.1% Triton X-100 permeabilized 3T3 cells.

HDAC Activity:
This antibody has been reported to immunoprecipitate active HDAC.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histones
Use Anti-HDAC1 Antibody (Rabbit Polyclonal Antibody) validated in EA, ICC, IP, WB to detect HDAC1 also known as histone deacetylase 1.

Quality

Routinely evaluated by western blot on RIPA lysates from 3T3/A31 cells.

Western Blot Analysis:
0.5-2 μg/mL of this lot detected HDAC1 in RIPA lysates from 3T3/A31 cells.

Target description

65 kDa

Physical form

Format: Purified
Protein A chromatography
Purified rabbit polyclonal IgG in buffer containing 0.1 M Tris-glycine, pH 7.4, 0.15 M NaCl, containing 0.05% sodium azide. Frozen at -20ºC.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.

Handling Recommendations:
Upon first thaw, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
Positive Antigen Control: Catalog #12-305, 3T3/A31 lysate. Add 2.5 μL of 2-mercapto-ethanol/100 μL of lysate and boil for 5 minutes to reduce the preparation. Load 20 μg of reduced lysate per lane for minigels.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

HERP, a novel heterodimer partner of HES/E(spl) in Notch signaling.
T Iso, V Sartorelli, C Poizat, S Iezzi, H Y Wu, G Chung, L Kedes, Y Hamamori
Molecular and cellular biology null
Mi-Sung Kim et al.
The Journal of clinical investigation, 126(11), 4372-4386 (2016-11-02)
Obese, insulin-resistant states are characterized by a paradoxical pathogenic condition in which the liver appears to be selectively insulin resistant. Specifically, insulin fails to suppress glucose production, yet successfully stimulates de novo lipogenesis. The mechanisms underlying this dysregulation remain controversial.
Histone deacetylase-associating Atrophin proteins are nuclear receptor corepressors.
Wang, L; Rajan, H; Pitman, JL; McKeown, M; Tsai, CC
Genes & Development null
Li Wang et al.
Journal of cellular biochemistry, 117(6), 1429-1438 (2015-11-14)
The expression of BRDT, a member of the BET sub-family of double bromodomain-containing proteins, is restricted to the male germ line, specifically to pachytene-diplotene spermatocytes and early spermatids. We previously showed that loss of the first bromodomain of BRDT by
Mariana Urrutia et al.
PloS one, 11(9), e0163735-e0163735 (2016-09-24)
The neurotransmitter GABA has been recently identified as a potent immunosuppressive agent that targets both innate and adaptive immune systems and prevents disease progression of several autoimmunity models. Mesenchymal stem cells (MSCs) are self-renewing progenitor cells that differentiate into various

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service