Skip to Content
Merck
All Photos(1)

Key Documents

06-680-AF555

Sigma-Aldrich

Anti-Androgen Receptor Antibody, Alexa Fluor 555 Conjugate

from rabbit, ALEXA FLUOR 555

Synonym(s):

Androgen receptor, Dihydrotestosterone receptor, Nuclear receptor subfamily 3 group C member 4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.45

biological source

rabbit

Quality Level

conjugate

ALEXA FLUOR 555

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, rat

species reactivity (predicted by homology)

mouse (based on 100% sequence homology), canine (based on 100% sequence homology), horse (based on 100% sequence homology), bovine (based on 100% sequence homology)

technique(s)

immunocytochemistry: suitable

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

bovine ... Ar(280675)
dog ... Ar(403588)
human ... AR(367)
mouse ... Ar(11835)
rat ... Ar(24208)

General description

Androgen receptor (UniProt P15207; also known as Dihydrotestosterone receptor, Nuclear receptor subfamily 3 group C member 4) is encoded by the Ar (also known as Andr, Nr3c4, Tfm) gene (Gene ID 24208) in rat species. Androgen receptor (AR) is a hormone-activated transcription regulator that plays important roles in normal prostate development and function, as well as in malignant prostate transformation. The binding of an androgenic hormone (testosterone or dihydrotestosterone) induces a conformational change in AR, resulting in its dissociation from heat schock proteins and subsequent translocation from the cytosol to nucleus, where it dimerizes and mediates either transcription activation or repression in a target gene-specific manner. The androgen insensitivity syndrome (AIS) or testicular feminization (TFM) is caused by a mutation of the androgen receptor gene located on the X chromosome.

Immunogen

Epitope: N-terminal modulating region.
KLH-conjugated linear peptide corresponding to the N-terminal sequence of rat androgen receptor.

Application

Research Category
Epigenetics & Nuclear Function
Research Sub Category
Hormones & Receptors
The unconjugated antibody (Cat. No. 06-680) is shown to be suitable also for chromatin immunoprecipitation (ChIP), ChIP-seq, Western blotting, immunoprecipitation, flow cytometry, immunohistochemistry applications.
This Anti-Androgen Receptor Antibody, Alexa Fluor 555 Conjugate is validated for use in Immunocytochemistry for the detection of Androgen Receptor.

Quality

Evaluated by Immunocytochemistry in T47D cells.

Immunocytochemistry Analysis: A 1:100 dilution of this antibody detected androgen receptor in T47D human breast cancer cells.

Target description

~110 kDa observed.

Physical form

Affinity purified.
Purified rabbit polyclonal antibody conjugate in PBS with 15 mg/mL BSA and 0.1 % sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Legal Information

ALEXA FLUOR is a trademark of Life Technologies

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Erik A Burlingame et al.
Cell reports methods, 1(4) (2021-09-07)
The emergence of megascale single-cell multiplex tissue imaging (MTI) datasets necessitates reproducible, scalable, and robust tools for cell phenotyping and spatial analysis. We developed open-source, graphics processing unit (GPU)-accelerated tools for intensity normalization, phenotyping, and microenvironment characterization. We deploy the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service