Skip to Content
Merck
All Photos(1)

Key Documents

05-253

Sigma-Aldrich

Anti-ZAP-70 Antibody, clone 2F3.2

clone 2F3.2, Upstate®, from mouse

Synonym(s):

Anti-ADMIO2, Anti-SRK, Anti-STCD, Anti-STD, Anti-TZK, Anti-ZAP-70

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2F3.2, monoclonal

species reactivity

human

manufacturer/tradename

Upstate®

technique(s)

flow cytometry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2a

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... ZAP70(7535)

General description

Note: This product is for research use only. It is not manufacured for diagnostic or therapeutic use in humans or animals.
Zeta-chain-associated protein kinase 70. A member of the protein tyrosine kinase family, ZAP-70 is normally expressed in T cells and natural killer cells and has a critical role in the initiation of T-cell signaling. ZAP-70 is expressed in T cells and tumors of T-cell lineage. A high level of ZAP-70 expression appears restricted to T-cell proliferative diseases and a subgroup of chronic lymphocytic leukemia (CLL). The ZAP-70 gene is in chromosome 2q12.

Specificity

Recognizes human ZAP-70, does not recognize murine ZAP-70.

Immunogen

GST-fusion to tandem SH2 domains of human ZAP-70 corresponding to residues 1-254

Application

For testing in IHC, please see the following link (PDF): http://www.propathlab.com/pdf/focus_dec_2003.pdf
Research Category
Signaling
Research Sub Category
Immunological Signaling
This Anti-ZAP-70 Antibody, clone 2F3.2 is validated for use in FC, IH, IP, WB for the detection of ZAP-70.

Quality

routinely evaluated on RIPA lysates prepared from Jurkat cells or Hut-78 cells

Target description

70 kDa

Physical form

Format: Purified
Protein G Purified
Protein G Purified immunoglobulin in Protein G Purified immunoglobulin in 30% glycerol, 0.07M Tris-glycine, pH 7.4, 0.105 M NaCl, 0.035% sodium azide as a preservative.

Storage and Stability

Maintain for 2 years at -20°C from date of shipment. Aliquot to avoid repeated freezing and thawing. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Analysis Note

Control
Positive Antigen Control: Catalog #12-303, Jurkat cell lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nir Shani et al.
Blood, 113(15), 3530-3541 (2008-10-22)
The default pathway of cell-surface T-cell receptor (TCR) complex formation, and the subsequent transport to the membrane, is thought to entail endoplasmic reticulum (ER) localization followed by proteasome degradation of the unassembled chains. We show herein an alternative pathway: short
Alexander Kröber et al.
Journal of clinical oncology : official journal of the American Society of Clinical Oncology, 24(6), 969-975 (2006-01-19)
Immunoglobulin heavy chain variable-region (VH) gene mutation status and zeta-associated protein 70 (ZAP-70) expression are correlated in chronic lymphocytic leukemia (CLL), but their concordance is variable. The goal of this study was to elucidate additional factors potentially characterizing their discordance.
C Silvin et al.
The Journal of biological chemistry, 276(24), 21450-21457 (2001-04-03)
Wiskott-Aldrich syndrome protein (WASP) plays a key role in cytoskeletal rearrangement and transcriptional activation in T-cells. Recent evidence links WASP and related proteins to actin polymerization by the Arp2/3 complex. To study whether the role of WASP in actin polymerization
Genetic evidence of a role for Lck in T-cell receptor function independent or downstream of ZAP-70/Syk protein tyrosine kinases
Wong, J., et al
Molecular and cellular biology, 18, 2855-2866 (1998)
Eva Calpe et al.
PloS one, 8(12), e81221-e81221 (2013-12-07)
ZAP-70 in chronic lymphocytic leukemia (CLL) is associated with enhanced response to microenvironmental stimuli. We analyzed the functional consequences of ZAP-70 ectopic expression in malignant B-cells in a xenograft mouse model of disseminated B-cell leukemia. Mice injected with B-cells expressing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service