Skip to Content
Merck
All Photos(1)

Key Documents

N-924

Supelco

Nalorphine hydrochloride solution

1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H21NO3·HCl
CAS Number:
Molecular Weight:
347.84
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

Cl.O[C@H]1C=C[C@H]2[C@H]3Cc4ccc(O)c5O[C@@H]1[C@]2(CCN3CC=C)c45

InChI

1S/C19H21NO3.ClH/c1-2-8-20-9-7-19-12-4-6-15(22)18(19)23-17-14(21)5-3-11(16(17)19)10-13(12)20;/h2-6,12-13,15,18,21-22H,1,7-10H2;1H/t12-,13+,15-,18-,19-;/m0./s1

InChI key

NAHATSPWSULUAA-ZQGPYOJVSA-N

General description

A certified solution standard applicable for use as starting material in calibrators or controls for a variety of LC/MS or GC/MS applications such as clinical toxicology or forensic analysis. Nalorphine is an opiate used to counteract the effects of opioid overdose.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Y Cheng et al.
Bioorganic & medicinal chemistry, 4(1), 73-80 (1996-01-01)
N-Cubylmethylnormorphine (1) and N-cubylmethylnoroxymorphone (2) have been synthesized and found to be more potent ligands at the mu and kappa opioid receptors than morphine and oxymorphone respectively. In the guinea-pig ileum preparation, compounds 1 and 2 were characterized as opioid
M R Bouw et al.
Pharmaceutical research, 15(11), 1673-1679 (1998-12-02)
To investigate the performance of two alternative retrodialysis recovery methods and to describe the influence of different recoveries on the reliability in estimating unbound extracellular concentrations of morphine. Unbound concentrations of morphine in striatum and in blood were determined by
C D King et al.
Drug metabolism and disposition: the biological fate of chemicals, 25(2), 251-255 (1997-02-01)
Opioids are important drugs used as analgesics, antitussives, antidiarrheals, and in the therapy of myocardial infarctions, and as antagonists of opioid intoxication. The glucuronidation of these compounds, catalyzed by UDP-glucuronosyltransferases (UGTs), is well known to be a primary step in
C D King et al.
Archives of biochemistry and biophysics, 332(1), 92-100 (1996-08-01)
Rat and human UDP-glucuronosyltransferase (UGT) 1.1 share > 70% identity in their deduced primary amino acid sequences. We have previously shown that rat UGT1.1, stably expressed in human embryonic kidney 293 cells, catalyzes the glucuronidation of bilirubin and the mixed
T N Decaestecker et al.
Rapid communications in mass spectrometry : RCM, 14(19), 1787-1792 (2000-09-28)
A liquid chromatographic mass spectrometric strategy for systematic toxicological analysis (STA) is presented using the automatic 'on-the-fly' single mass spectrometry mode to tandem mass spectrometry mode (MS to MS/MS) switching abilities of a quadrupole time-of-flight (Q-TOF) instrument. During the chromatographic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service