Skip to Content
MilliporeSigma

D-080

Demoxepam-d5 solution

100 μg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C15D5H6ClN2O2
Molecular Weight:
291.74
UNSPSC Code:
41116107
MDL number:
NACRES:
NA.24
EC Number:
200-835-2
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

Quality Segment

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule B (Switzerland)

concentration

100 μg/mL in acetonitrile

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

[2H]c1c([2H])c([2H])c(c([2H])c1[2H])C2=[N+]([O-])CC(=O)Nc3ccc(Cl)cc23

InChI

1S/C15H11ClN2O2/c16-11-6-7-13-12(8-11)15(10-4-2-1-3-5-10)18(20)9-14(19)17-13/h1-8H,9H2,(H,17,19)/i1D,2D,3D,4D,5D

InChI key

GGRWZBVSUZZMKS-RALIUCGRSA-N

General description

An internal standard suitable for GC/MS or LC/MS applications with chlordiazepoxide or demoxepam such as toxicology analysis, forensic testing, pain prescription monitoring, or isotope dilution methods. Demoxepam is a benzodiazepine drug and anticonvulsant as well as a primary metabolite of chlordiazepoxide, marketed as Klopoxid or Librax® for the treatment of anxiety and muscle spasms or pain.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Librax is a registered trademark of Valeant Pharmaceuticals North America LLC
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany


pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_f

35.6 °F - closed cup

flash_point_c

2 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library