Skip to Content
Merck

Skip To

D-072

Dopamine-D4 hydrochloride solution

100 μg/mL in methanol with 5% 1 M HCl (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

Dopamine-d4 hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C8H7D4NO2·HCl
CAS Number:
Molecular Weight:
193.66
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

Quality Segment

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol with 5% 1 M HCl (as free base)

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−70°C

SMILES string

Cl.NC(C(c1cc(c(cc1)O)O)([2H])[2H])([2H])[2H]

InChI

1S/C8H11NO2.ClH/c9-4-3-6-1-2-7(10)8(11)5-6;/h1-2,5,10-11H,3-4,9H2;1H/i3D2,4D2;

InChI key

CTENFNNZBMHDDG-URZLSVTISA-N

General description

A stable labeled internal standard suitable for the quantitation of dopamine levels in LC/MS applications such as diagnostic testing, endocrinology, and clinical chemistry. Dopamine is a catecholamine which acts as a neurotransmitter for dopamine receptors and neurohormone for regulation of numerous brain functions.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
C-113D-081C-091
application(s)

clinical testing

application(s)

clinical testing

application(s)

clinical testing

application(s)

clinical testing

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

form

liquid

form

liquid

form

liquid

form

liquid

Quality Level

300

Quality Level

300

Quality Level

300

Quality Level

300

grade

certified reference material

grade

certified reference material

grade

certified reference material

grade

certified reference material

storage temp.

−70°C

storage temp.

−20°C

storage temp.

−70°C

storage temp.

−20°C


signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes,Central nervous system

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_f

51.8 °F - closed cup

flash_point_c

11 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Weijie Ouyang et al.
Investigative ophthalmology & visual science, 62(1), 25-25 (2021-01-26)
To evaluate the role of CD4+ T helper cells in benzalkonium chloride (BAC)-induced ocular surface disorder in C57BL/6 mice. Topical 0.075% BAC was applied twice daily in C57BL/6 mice for 7 consecutive days; PBS-treated and untreated mice served as controls.



Global Trade Item Number

SKUGTIN
D-072-1ML04061833556795

Questions

Reviews

No rating value

Active Filters