Skip to Content
Merck
All Photos(2)

Key Documents

850482P

Avanti

16:0-18:0 (11-12BR) PC

1-palmitoyl-2-(11,12-dibromo)stearoyl-sn-glycero-3-phosphocholine, powder

Synonym(s):

1-hexadecanoyl-2-(11,12-dibromooctadecanoyl)-sn-glycero-3-phosphocholine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H82NO8PBr2
CAS Number:
Molecular Weight:
919.88
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 10 mg (850482P-10mg)

manufacturer/tradename

Avanti Research - A Croda Brand 850482P

lipid type

phospholipids
omega FAs

shipped in

dry ice

storage temp.

−20°C

SMILES string

[O-]P(OCC[N+](C)(C)C)(OC[C@]([H])(OC(CCCCCCCCCC(Br)C(Br)CCCCCC)=O)COC(CCCCCCCCCCCCCCC)=O)=O

InChI

1S/C42H82Br2NO8P/c1-6-8-10-12-13-14-15-16-17-18-21-24-28-32-41(46)50-36-38(37-52-54(48,49)51-35-34-45(3,4)5)53-42(47)33-29-25-22-19-20-23-27-31-40(44)39(43)30-26-11-9-7-2/h38-40H,6-37H2,1-5H3/t38-,39?,40?/m1/s1

InChI key

MPQPFPFIGLVSQR-YXGYHMDESA-N

General description

16:0-18:0 (11-12BR) PC (1-palmitoyl-2-(11,12-dibromo) stearoyl-sn-glycero-3-phosphocholine) is a brominated-PC lipid which has palmitoyl and stearoyl chains.

Application

16:0-18:0 (11-12BR) PC (1-palmitoyl-2-(11,12-dibromo) stearoyl-sn-glycero-3-phosphocholine) has been used in large unilamellar vesicle preparation. It may be used:
  • in phospholipid vesicle preparation for tryptophan (Trp) penetration depth experiments
  • to determine the bromination effect at different positions along the fatty acid chains
  • in large unilamellar vesicle (LUV) for quenching of human heat shock protein 70 (Hsp70) tryptophan (Trp) fluorescence

Biochem/physiol Actions

16:0-18:0 (11-12BR) PC (1-palmitoyl-2-(11,12-dibromo) stearoyl-sn-glycero-3-phosphocholine) can be used to prepare large unilamellar vesicles.

Packaging

5 mL Clear Glass Sealed Ampule (850482P-10mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ajay K Mahalka et al.
Biochimica et biophysica acta, 1838(5), 1344-1361 (2014-02-01)
While a significant fraction of heat shock protein 70 (Hsp70) is membrane associated in lysosomes, mitochondria, and the outer surface of cancer cells, the mechanisms of interaction have remained elusive, with no conclusive demonstration of a protein receptor. Hsp70 contains
Henriette Aksnes et al.
The Journal of biological chemistry, 292(16), 6821-6837 (2017-02-16)
Nα-Acetyltransferase 60 (Naa60 or NatF) was recently identified as an unconventional N-terminal acetyltransferase (NAT) because it localizes to organelles, in particular the Golgi apparatus, and has a preference for acetylating N termini of the transmembrane proteins. This knowledge challenged the
Tim Rasmussen et al.
Journal of molecular biology, 431(17), 3339-3352 (2019-06-08)
All membrane proteins have dynamic and intimate relationships with the lipids of the bilayer that may determine their activity. Mechanosensitive channels sense tension through their interaction with the lipids of the membrane. We have proposed a mechanism for the bacterial
Candace M Pfefferkorn et al.
Biochimica et biophysica acta, 1848(11 Pt A), 2821-2828 (2015-08-25)
Apolipoproteins are essential human proteins for lipid metabolism. Together with phospholipids, they constitute lipoproteins, nm to μm sized particles responsible for transporting cholesterol and triglycerides throughout the body. To investigate specific protein-lipid interactions, we produced and characterized three single-Trp containing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service