Skip to Content
Merck
All Photos(2)

Documents

840031P

Avanti

08:0 PS

1,2-dioctanoyl-sn-glycero-3-phospho-L-serine (sodium salt), powder

Synonym(s):

PS(8:0/8:0)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H41NO10PNa
CAS Number:
Molecular Weight:
533.53
UNSPSC Code:
51191904
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 10 mg (840031P-10mg)
pkg of 1 × 200 mg (840031P-200mg)

manufacturer/tradename

Avanti Research - A Croda Brand 840031P

lipid type

phospholipids
cardiolipins

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OC[C@](C([O-])=O)([H])[NH3+])=O)(OC(CCCCCCC)=O)COC(CCCCCCC)=O.[Na+]

InChI

1S/C22H42NO10P.Na/c1-3-5-7-9-11-13-20(24)30-15-18(33-21(25)14-12-10-8-6-4-2)16-31-34(28,29)32-17-19(23)22(26)27;/h18-19H,3-17,23H2,1-2H3,(H,26,27)(H,28,29);/q;+1/p-1/t18-,19+;/m1./s1

InChI key

IOFLNDDTVDTSHA-VOMIJIAVSA-M

General description

Phosphatidylserine (PS), an acidic phospholipid is obtained from soybeans (soy-PS), bovine cortex (BC-PS) and synthetic derivation. It is mainly seen in animals, higher plants and microorganisms. It is localized in the inner leaflet of the plasma membrane.

Application

08:0 PS may be used as control anionic lipid to treat Xenopus eggs to study its effects on the fertilization events. It has also been used to form mixed micelles to study or compare the micellization behavior of mixed zwitterionic/cationic solutions and cationic gemini surfactants with that of 08:0 PS.

Biochem/physiol Actions

Phosphatidylserine (PS) can activate protein kinase C. It aids in removing apoptotic cells and also participates in platelet aggregation.

Packaging

20 mL Clear Glass Screw Cap Vial (840031P-200mg)
5 mL Clear Glass Sealed Ampule (840031P-10mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Analysis of complex lipidomes
Medical Applications of Mass Spectrometry, 223-249 (2008)
Effect of surfactant structure on the mixed micelle formation of cationic gemini-zwitterionic phospholipid systems
Naqvi AZ, et al.
Colloids and Surfaces. A, Physicochemical and Engineering Aspects, 477 (2015)
Cognitive Effects of Nutraceuticals
Nutraceuticals, 29-48 (2016)
Analysis of complex lipidomes
Medical Applications of Mass Spectrometry, 223-249 (2008)
Ryan C Bates et al.
Developmental biology, 386(1), 165-180 (2013-11-26)
We report a new step in the fertilization in Xenopus laevis which has been found to involve activation of Src tyrosine kinase to stimulate phospholipase C-γ (PLC-γ) which increases inositol 1,4,5-trisphosphate (IP3) to release intracellular calcium ([Ca](i)). Molecular species analysis

Articles

The critical micelle concentration (CMC) can be approximately defined as the lipid monomer concentration at which appreciable amounts (>5% of total) of micellar aggregates first begin to appear in the equilibrium: nM1<=>Mn

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service