Skip to Content
Merck
All Photos(2)

Key Documents

700062P

Avanti

stigmasterol

Avanti Research - A Croda Brand

Synonym(s):

Stigmasterin; Phytosterol; beta-Stigmasterol; D5-Stigmasterol; 110801

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H48O
CAS Number:
Molecular Weight:
412.69
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25
Pricing and availability is not currently available.

description

stigmasta-5,22-dien-3-ol

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 5 mg (700062P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@H]1CC[C@@]2([C@@H]3[C@H]([C@H]4[C@@]([C@H](CC4)[C@H](C)\C=C\[C@H](C(C)C)CC)(CC3)C)CC=C2C1)C

InChI

1S/C29H48O/c1-7-21(19(2)3)9-8-20(4)25-12-13-26-24-11-10-22-18-23(30)14-16-28(22,5)27(24)15-17-29(25,26)6/h8-10,19-21,23-27,30H,7,11-18H2,1-6H3/b9-8+/t20-,21-,23+,24+,25-,26+,27+,28+,29-/m1/s1

InChI key

HCXVJBMSMIARIN-PHZDYDNGSA-N

General description

Stigmasterol is a plant sterol that comprises an unsaturated bond between C22 and C23. It is found in foods like margarines and yogurts.[1] Stigmasterol is synthesized from the mevalonate pathway and is present during development stages in plants.[2]

Application

Stigmasterol has been used as a phytosterol compound to test its anti-proliferative property in breast cancer cell culture and its effect on liver X receptor-α (LXRA) activation.[1] It may be used as an internal standard for the quantification of yeast cell wall sterols using gas chromatography–mass spectrometry (GC-MS) analysis,[3] and in the preparation of lipid monolayers.[4]

Biochem/physiol Actions

Stigmasterol possesses anti-inflammatory anti-hypercholestrolemic, antitumor and antioxidant functionality.[5] It plays a crucial role in the activation of plasma membrane H+-ATPase and cell proliferation. Variations in stigmasterol and its precursor are noticeable at both the seed and whole plant developmental stages. Stigmasterol may be involved in gravitropism and tolerance to abiotic stress.[2]

Packaging

5 mL Amber Glass Screw Cap Vial (700062P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stigmasterol: a comprehensive review
Kaur N, et al.
International Journal of Pharmaceutical Sciences and Research, 2(9), 2259-2259 (2011)
Larissa V G Longo et al.
PloS one, 8(5), e63372-e63372 (2013-05-22)
The fungal cell wall is a complex and dynamic outer structure. In pathogenic fungi its components interact with the host, determining the infection fate. The present work aimed to characterize cell wall lipids from P. brasiliensis grown in the presence
Katarzyna Hąc-Wydro et al.
Colloids and surfaces. B, Biointerfaces, 116, 138-146 (2014-01-28)
In this work miscibility and interactions of sterols with choline plasmalogen (PC-plasm) in Langmuir monolayers were studied. Moreover, the properties of cholesterol/phosphatidylcholine/plasmalogen mixtures of different PC-plasm concentration were investigated. The foregoing systems were treated as a model of cancer cell
Siddique I Aboobucker et al.
Frontiers in plant science, 10, 354-354 (2019-04-16)
A direct role for cholesterol signaling in mammals is clearly established; yet, the direct role in signaling for a plant sterol or sterol precursor is unclear. Fluctuations in sitosterol and stigmasterol levels during development and stress conditions suggest their involvement
Samantha A Hutchinson et al.
International journal of molecular sciences, 20(13) (2019-07-05)
Low fruit and vegetable consumption and high saturated fat consumption causes elevated circulating cholesterol and are breast cancer risk factors. During cholesterol metabolism, oxysterols form that bind and activate the liver X receptors (LXRs). Oxysterols halt breast cancer cell proliferation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service