Key Documents
Y907227
Sigma-Aldrich
Valve
brass diaphragm, for CGA 350 right angle path, external, left hand thread
About This Item
UNSPSC Code:
12142200
NACRES:
NA.12
material
brass diaphragm
Quality Level
parameter
2640 psi max. pressure
L
4.62 in.
weight
613 g
fitting
for CGA 350 right angle path, external, left hand thread
Other Notes
This product is only available with the purchase of a bulk gas product and not for individual sale. For more information, please contact Stable Isotopes Customer Service
Choose from one of the most recent versions:
Certificates of Analysis (COA)