Skip to Content
Merck
All Photos(1)

Documents

W421501

Sigma-Aldrich

Tyramine

98%, FG

Synonym(s):

2-(4-Hydroxyphenyl)ethylamine, 4-(2-Aminoethyl)phenol, 4-Hydroxyphenethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H4CH2CH2NH2
CAS Number:
Molecular Weight:
137.18
FEMA Number:
4215
Beilstein:
1099914
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
11.007
NACRES:
NA.21

biological source

synthetic

grade

FG

reg. compliance

EU Regulation 1334/2008 & 872/2012
FDA 21 CFR 110

Assay

98%

bp

175-181 °C/8 mmHg (lit.)

mp

160-162 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

meaty; phenolic

SMILES string

NCCc1ccc(O)cc1

InChI

1S/C8H11NO/c9-6-5-7-1-3-8(10)4-2-7/h1-4,10H,5-6,9H2

InChI key

DZGWFCGJZKJUFP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Can enter catecholaminergic terminals and be released as a false transmitter.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stephan A Chalon et al.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology, 28(9), 1685-1693 (2003-06-05)
Evidence suggests that compounds that increase the synaptic availability of more than one neurotransmitter have greater efficacy in the treatment of depression than single-acting drugs. Preclinical studies indicate that duloxetine acts to inhibit serotonin (5-HT) and norepinephrine (NE) transporters. The
Giris Jacob et al.
Circulation, 107(19), 2475-2479 (2003-04-23)
Despite the widespread use of tyramine as a pharmacological tool to assess the effects of norepinephrine release from sympathetic nerve terminals, its vascular effects are not adequately characterized. In particular, previous results indicate that intravenous tyramine produces little if any
D R Varma et al.
The Journal of pharmacology and experimental therapeutics, 265(3), 1096-1104 (1993-06-01)
At concentrations higher than that required to produce maximal vasoconstriction, tyramine caused concentration-dependent relaxation of rat aortic strips contracted maximally by tyramine, norepinephrine, phenylephrine, 5-hydroxytryptamine, prostaglandin F2 alpha, endothelin, angiotensin II and potassium; isoproterenol did not relax potassium-contracted strips. The
Thomas Roeder
Annual review of entomology, 50, 447-477 (2004-09-10)
Octopamine (OA) and tyramine (TA) are the invertebrate counterparts of the vertebrate adrenergic transmitters. They are decarboxylation products of the amino acid tyrosine, with TA as the biological precursor of OA. Nevertheless, both compounds are independent neurotransmitters that act through
Adam B Braunschweig et al.
Nano letters, 7(7), 2030-2036 (2007-06-15)
Tyrosinase activity is monitored by pi-donor-acceptor force interactions between a bipyridinium-modified AFM tip and the biocatalytic reaction product generated on a tyramine- (or dopamine-) modified surface. Upon oxidation of the surface to dopaquinone as a result of tyrosinase activity, force

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service