Skip to Content
Merck
All Photos(1)

Key Documents

W319600

Sigma-Aldrich

cis-Jasmone

stabilized

Synonym(s):

Jasmone, cis-3-Methyl-2-(2-pentenyl)-2-cyclopenten-1-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H16O
CAS Number:
Molecular Weight:
164.24
FEMA Number:
3196
Beilstein:
1907713
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
7.094
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
FDA 21 CFR 172.515

contains

α-tocopherol as stabilizer

refractive index

n20/D 1.498 (lit.)

bp

134-135 °C/12 mmHg (lit.)

density

0.94 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

woody; floral; herbaceous

SMILES string

[H]\C(CC)=C(/[H])CC1=C(C)CCC1=O

InChI

1S/C11H16O/c1-3-4-5-6-10-9(2)7-8-11(10)12/h4-5H,3,6-8H2,1-2H3/b5-4-

InChI key

XMLSXPIVAXONDL-PLNGDYQASA-N

Looking for similar products? Visit Product Comparison Guide

General description

cis-Jasmone is one of the key volatile flavor compounds in different kinds of tea such as green tea, Oolong tea, jasmine tea and Tie Guanyin tea. It is also the main odor constituent of jasmine oil. In plants, cis-Jasmone acts as an activator of defense during an insect attack or mechanical damage.

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

224.6 °F - closed cup

Flash Point(C)

107 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Photochemical reaction of (Z)-jasmone under various conditions.
Tateba H, et al.
Bioscience, Biotechnology, and Biochemistry, 57(2), 220-226 (1993)
Flavor constituents of pouchong tea and a comparison of the aroma pattern with jasmine tea
Yamanishi T, et al.
Agricultural and Biological Chemistry, 44(9), 2139-2142 (1980)
New roles for cis-jasmone as an insect semiochemical and in plant defense.
Birkett MA, et al.
Proceedings of the National Academy of Sciences of the USA, 97(16), 9329-9334 (2000)
Preparation of the volatile flavor components from Tie Guanyin Tea and its application in cigarette flavoring.
Yuexin NI
Flavour Fragrance Cosmetics, 3, 416-416 (2012)
Effect of thermal treatment on the flavor formation of Oolong tea.
Yu TH, et al.
Food science and agricultural chemistry (1999)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service