Skip to Content
Merck
All Photos(1)

Key Documents

W250902

Sigma-Aldrich

Geranyl acetate

FCC

Synonym(s):

trans-3,7-Dimethyl-2,6-octadien-1-yl acetate, trans-3,7-Dimethyl-2,6-octadienyl acetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CO2CH2CH=C(CH3)CH2CH2CH=C(CH3)2
CAS Number:
Molecular Weight:
196.29
FEMA Number:
2509
EC Number:
Council of Europe no.:
201
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.011
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

Halal
Kosher

reg. compliance

FCC
FDA 21 CFR 117

vapor density

6.8 (vs air)

vapor pressure

0.07 mmHg ( 20 °C)

refractive index

n20/D 1.461 (lit.)

bp

138 °C/25 mmHg (lit.)

density

0.907 g/cm3 at 25 °C

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

fruity; rose; sweet

SMILES string

CC(=O)OC\C=C(/C)CC\C=C(/C)C

InChI

1S/C12H20O2/c1-10(2)6-5-7-11(3)8-9-14-12(4)13/h6,8H,5,7,9H2,1-4H3/b11-8+

InChI key

HIGQPQRQIQDZMP-DHZHZOJOSA-N

Looking for similar products? Visit Product Comparison Guide

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

219.2 °F - closed cup

Flash Point(C)

104 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Satoshi Nojima et al.
PloS one, 6(3), e18178-e18178 (2011-04-06)
Semiochemicals are often produced in infinitesimally small quantities, so their isolation requires large amounts of starting material, not only requiring significant effort in sample preparation, but also resulting in a complex mixture of compounds from which the bioactive compound needs
Won-Il Cho et al.
Journal of food protection, 78(6), 1221-1225 (2015-06-04)
The sporicidal activities against Bacillus subtilis spores of surfactant components with hydrophilic and hydrophobic properties that can lead to the denaturation of various proteins comprising the spore structure were investigated. The reduction in spore numbers by each of the surfactant
Bio-degradation of acetates of geraniol, nerol & citronellol by P. incognita: isolation & identification of metabolites.
K M Madyastha et al.
Indian journal of biochemistry & biophysics, 20(3), 136-140 (1983-06-01)
Luca Nicolotti et al.
Journal of chromatography. A, 1360, 264-274 (2014-08-19)
Comprehensive two-dimensional gas chromatography (GC×GC) coupled with Mass Spectrometry (MS) is one of today's most powerful analytical platforms for detailed analysis of medium-to-high complexity samples. The column set usually consists of a long, conventional-inner-diameter first dimension ((1)D) (typically 15-30m long
Kye Chung Park et al.
Journal of insect physiology, 78, 33-46 (2015-05-06)
Phytophagous insects detect volatile compounds produced by host and non-host plants, using species-specific sets of olfactory receptor neurons (ORNs). To investigate the relationship between the range of host plants and the profile of ORNs, single sensillum recordings were carried out

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service