Skip to Content
Merck
All Photos(1)

Key Documents

W246409

Sigma-Aldrich

Ethyl vanillin

≥98%, FCC, FG

Synonym(s):

3-Ethoxy-4-hydroxybenzaldehyde, Ethylvanillin, NSC 1803, NSC 67240

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5OC6H3(OH)CHO
CAS Number:
Molecular Weight:
166.17
FEMA Number:
2464
Beilstein:
1073761
EC Number:
Council of Europe no.:
108
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
5.019
NACRES:
NA.21
Organoleptic:
creamy; sweet; vanilla
grade:
FG
Halal
Kosher
biological source:
synthetic
Agency:
meets purity specifications of JECFA
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 117
FDA 21 CFR 182.60
FDA 21 CFR 182.90

vapor pressure

<0.01 mmHg ( 25 °C)

Assay

≥98%

form

solid

mp

74-77 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

creamy; sweet; vanilla

SMILES string

[H]C(=O)c1ccc(O)c(OCC)c1

InChI

1S/C9H10O3/c1-2-12-9-5-7(6-10)3-4-8(9)11/h3-6,11H,2H2,1H3

InChI key

CBOQJANXLMLOSS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethyl vanillin is one of the main components of artificial vanilla flavorings. It is also used a flavor enhancer.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

293.0 °F - closed cup

Flash Point(C)

145 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kakuyou Ogawa et al.
Journal of natural medicines, 72(3), 798-802 (2018-03-24)
Vanilla flavour is familiar to consumers through foods, cosmetics, household products and some medicines. Vanilla flavouring agents typically contain vanillin or its analogue ethyl vanillin. Our previous study revealed that the inhalation of eugenol, which contains a vanillyl group, has
Control of undeclared flavoring of cocoa powders by the determination of vanillin and ethyl vanillin by HPLC.
Perez-Esteve E, et al.
Food Control, 67, 171-176 (2016)
Microencapsulation of flavors in carnauba wax.
Milanovic J, et al.
Sensors, 10(1), 901-912 (2010)
Laila Ali et al.
Journal of AOAC International, 91(2), 383-386 (2008-05-15)
A method is described for determining coumarin, vanillin, and ethyl vanillin in vanilla extract products. A product is diluted one-thousand-fold and then analyzed by reversed-phase liquid chromatography using a C18 column and a mobile phase consisting of 55% acetonitrile-45% aqueous
Tadashi Ishimaru
The Annals of otology, rhinology, and laryngology, 116(5), 335-341 (2007-06-15)
Endoscopy of the human olfactory cleft is important for both research in human olfaction and clinical examination with regard to olfactory disorders. However, endoscopy only provides information on the morphology and functional status of the epithelium, and it does not

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service