Skip to Content
Merck
All Photos(1)

Key Documents

W215015

Sigma-Aldrich

Benzyl propionate

natural, ≥98%, FCC, FG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5CO2CH2C6H5
CAS Number:
Molecular Weight:
164.20
FEMA Number:
2150
EC Number:
Council of Europe no.:
413
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.132
NACRES:
NA.21

grade

FG
Fragrance grade
Kosher
natural

Quality Level

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 117

Assay

≥98%

refractive index

n20/D 1.497 (lit.)

bp

222 °C (lit.)

density

1.03 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

jasmine; floral; fruity; sweet

SMILES string

CCC(=O)OCc1ccccc1

InChI

1S/C10H12O2/c1-2-10(11)12-8-9-6-4-3-5-7-9/h3-7H,2,8H2,1H3

InChI key

VHOMAPWVLKRQAZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

204.8 °F - closed cup

Flash Point(C)

96 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service