All Photos(2)
About This Item
Linear Formula:
C6H5C6H4C6H5
CAS Number:
Molecular Weight:
230.30
Beilstein:
1864778
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
99%
bp
379 °C (lit.)
mp
84-88 °C (lit.)
SMILES string
c1ccc(cc1)-c2cccc(c2)-c3ccccc3
InChI
1S/C18H14/c1-3-8-15(9-4-1)17-12-7-13-18(14-17)16-10-5-2-6-11-16/h1-14H
InChI key
YJTKZCDBKVTVBY-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
- Construction of double-stranded metallosupramolecular polymers with a controlled helicity by combination of salt bridges and metal coordination.: Describes the construction of metallosupramolecular polymers, integrating m-Terphenyl derivatives for controlled helical structures, significant for developing responsive materials (Ikeda et al., 2006).
- A new class of oxo-bridged high-valent hexamanganese clusters supported by sterically hindered carboxylate ligands.: Focuses on a new class of manganese clusters utilizing m-Terphenyl-based carboxylate ligands, relevant for magnetic and catalytic applications (Mukhopadhyay et al., 2003).
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Acute 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Christin Suenkel et al.
Cell reports, 30(7), 2170-2179 (2020-02-23)
circSLC45A4 is the main RNA splice isoform produced from its genetic locus and one of the highest expressed circRNAs in the developing human frontal cortex. Knockdown of this highly conserved circRNA in a human neuroblastoma cell line is sufficient to induce
T S Mayadevi et al.
Membranes, 10(11) (2020-11-11)
A series of poly(meta/para-terphenylene-methyl piperidinium)-based anion exchange membranes devoid of benzylic sites or aryl ether bonds, that are vulnerable to degradation by hydroxide ions, are synthesized and investigated for their application as novel anion exchange membranes. The copolymers are composed
Shijia Liu et al.
Analytical and bioanalytical chemistry, 407(30), 8989-8997 (2015-10-03)
A novel imidazolium-embedded N,N-dimethylaminopropyl-functionalized silica-based stationary phase (Sil-ImCl) was prepared and further used for hydrophilic interaction/reversed-phase mixed-mode chromatography. The Sil-ImCl stationary phase was respectively characterized by Fourier transform infrared spectrometry, thermogravimetric analysis, and element analysis. A variety of hydrophilic or
Lijuan Luo et al.
Scientific reports, 5, 12776-12776 (2015-08-05)
Algal blooms cause great damage to water quality and aquaculture. However, this study showed that dead algal cells and chlorophyll could accelerate the photo-transformation of benzo[a]pyrene (BaP), a ubiquitous and persistent pollutant with potently mutagenic and carcinogenic toxicities, under visible
Xiaotian Zhang et al.
Environmental science & technology, 49(16), 10127-10135 (2015-07-23)
Suspended sediment (SPS) is widely present in rivers around the world. However, the bioavailability of hydrophobic organic compounds (HOCs) associated with SPS is not well understood. In this work, the influence of SPS grain size on the bioavailability of SPS-associated
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service