SYNF352440M
Synthware™ fritted filter tube with glass stopcocks
joint: ST/NS 24/40, disc O.D. 36 mm, porosity: medium
Synonym(s):
Synthware F352440M, Filter tubes with fritted disc
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
Recommended Products
material
colorless tube
glass tube
Joint
joint: ST/NS24/40
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F352440M
disc O.D.
36 mm
General description
With two 2 mm glass stopcocks having constant pressure retention devices.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)