Skip to Content
Merck
All Photos(2)

Documents

P8135

Sigma-Aldrich

Sodium pyrophosphate dibasic

practical grade

Synonym(s):

Disodium dihydrogen pyrophosphate, Sodium diphosphate dibasic

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2H2P2O7
CAS Number:
Molecular Weight:
221.94
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

grade

practical grade

Assay

≥90% NaOH basis (titration)

form

powder

SMILES string

[Na+].[Na+].OP([O-])(=O)OP(O)([O-])=O

InChI

1S/2Na.H4O7P2/c;;1-8(2,3)7-9(4,5)6/h;;(H2,1,2,3)(H2,4,5,6)/q2*+1;/p-2

InChI key

GYQBBRRVRKFJRG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Sodium pyrophosphate dibasic can be used as a heavymetal extractant from sludge.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Caroline Öhman et al.
Journal of materials science. Materials in medicine, 26(7), 210-210 (2015-07-15)
The porosity of calcium phosphate cements has an impact on several important parameters, such as strength, resorbability and bioactivity. A model to predict the porosity for biomedical cements would hence be a useful tool. At the moment such a model
Bruce L Riser et al.
Nephrology, dialysis, transplantation : official publication of the European Dialysis and Transplant Association - European Renal Association, 26(10), 3349-3357 (2011-03-15)
The high rate of cardiovascular mortality in patients with end-stage renal disease (ESRD) is a significant barrier to improved life expectancy. Unique in this population is the marked development and aggressive worsening of vascular calcification (VC). Pyrophosphate (PPi), an endogenous
Priyadip Das et al.
Chemical communications (Cambridge, England), 47(28), 8118-8120 (2011-06-21)
Newly synthesized Zn(II) and Cd(II)-based complexes show unique selectivity towards inorganic pyrophosphate (PPi) in 100% aqueous medium at pH = 7.4 and act as a "turn-on" and "turn-off" real-time assay, respectively, for the evaluation of the enzymatic activity of alkaline
Taís Scaramucci et al.
Journal of dentistry, 39(12), 841-848 (2011-09-29)
To evaluate the erosive potential of orange juice modified by food additives in enamel and dentine. Calcium lactate pentahydrate (CLP), xanthan gum (XG), sodium linear polyphosphate (LPP), sodium pyrophosphate tetrabasic (PP), sodium tripolyphosphate (STP) and some of their combinations were
Nadia Marino et al.
Inorganic chemistry, 50(1), 378-389 (2010-12-01)
The reaction in water of Cu(NO(3))(2)·2.5H(2)O with 2,2'-bipyridine (bipy), 1,10-phenanthroline (phen), or 1,10-phenanthroline-5-amine (phenam), and sodium pyrophosphate (Na(4)P(2)O(7)), at various pHs, afforded three new copper(II)-pyrophosphate complexes, namely, {[Cu(bipy)(cis-H(2)P(2)O(7))](2)}·3H(2)O (1a), {[Cu(phen)(H(2)O)](4)(HP(2)O(7))(2)}(ClO(4))(2)·4H(2)O (2), and {[Cu(2)(phenam)(2)(P(2)O(7))](2)·25H(2)O}(n) (3). A solvent free crystalline phase of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service