Skip to Content
Merck
All Photos(2)

Key Documents

N109

Sigma-Aldrich

1-Naphthaldehyde

95%

Synonym(s):

α-Naphthal, Naphthalene-1-carbaldehyde, α-Naphthaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H7CHO
CAS Number:
Molecular Weight:
156.18
Beilstein:
386082
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

>1 (vs air)

Assay

95%

form

liquid

refractive index

n20/D 1.652 (lit.)

bp

160-161 °C/15 mmHg (lit.)

mp

1-2 °C (lit.)

density

1.15 g/mL at 25 °C (lit.)

SMILES string

[H]C(=O)c1cccc2ccccc12

InChI

1S/C11H8O/c12-8-10-6-3-5-9-4-1-2-7-11(9)10/h1-8H

InChI key

SQAINHDHICKHLX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1-Naphthaldehyde can be used to synthesize:
  • (S)-1-α-naphthyl-1-ethanol
  • N-(4-aryl)-N-(α-naphthyliden)amines
  • N-aryl-N-[1-(1-naphthyl)but-3-enyl]amines
  • naphthalene-1-carboxylic acid methyl ester

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Transformation of schiff bases derived from alpha?naphthaldehyde. Synthesis, spectral data and biological activity of new?3?aryl?2?(??naphtyl)?4?thiazolidinones and N?aryl?N?[1?(??naphthyl) but?3?enyl] amines.
Kouznetsov V, et al.
Journal of Heterocyclic Chemistry, 41(6), 995-999 (2004)
Miraç Ocak et al.
Journal of fluorescence, 27(1), 59-68 (2016-09-17)
Four new Schiff base ligands carrying naphthalene groups were prepared from the reaction of 2,4-diamino-6-methyl-1,3,5-triazine and 2,4-diamino-6-undecyl-1,3,5-triazine with 2-hydroxy-1-naphthaldehyde. The influence of a series of metal ions including Cu
Cytotoxic and Antifungal Activities of Diverse ?-Naphthylamine Derivatives.
Kouznetsov V, et al.
Scientia Pharmaceutica, 80(4), 867-878 (2012)
Pramod Kumar Singh et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 64(4), 853-858 (2006-03-15)
A series of new coordination complexes of cobalt(II), nickel(II) and copper(II) with two new aroylhydrazones, 2-hydroxy-1-naphthaldehyde isonicotinoylhydrazone (H(2)L(1)) and 2-hydroxy-1-naphthaldehyde-2-thenoyl-hydrazone (H(2)L(2)) have been synthesized and characterized by elemental analysis, conductance measurements, magnetic susceptibility measurements, (1)H NMR spectroscopy, IR spectroscopy, electronic
Highly enantioselective addition of dimethylzinc to arylaldehydes catalyzed by (2S)-1-ferrocenyl-methylaziridin-2-yl (diphenyl) methanol.
Wang M, et al.
Tetrahedron Asymmetry, 20(3), 288-292 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service