Skip to Content
Merck
All Photos(3)

Key Documents

M46845

Sigma-Aldrich

2-Methylfuran

contains 200-400 BHT as stabilizer, 99%

Synonym(s):

Silvan

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H6O
CAS Number:
Molecular Weight:
82.10
Beilstein:
103733
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

2.8 (vs air)

vapor pressure

139 mmHg ( 20 °C)
561 mmHg ( 55 °C)

Assay

99%

form

liquid

contains

200-400 BHT as stabilizer

refractive index

n20/D 1.433 (lit.)

bp

63-66 °C (lit.)

density

0.91 g/mL at 25 °C (lit.)

SMILES string

Cc1ccco1

InChI

1S/C5H6O/c1-5-3-2-4-6-5/h2-4H,1H3

InChI key

VQKFNUFAXTZWDK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Methylfuran, a substituted furan, is considered as a potential alternative fuel for internal combustion engines. Its microwave spectrum has been analyzed. The synthesis of 2-MF from carbon sources under different reaction conditions have been reported.

Application

2-Methylfuran may be used in the synthesis of exo-cis-1-methyl-7-oxabicyclo[2.2.1]hept-5-ene-2,3-dicarboxylic anhydride via Diels-Alder reaction with maleic anhydride.

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-7.6 °F

Flash Point(C)

-22 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Combustion performance and emissions of 2-methylfuran diesel blends in a diesel engine.
Xiao H, et al.
Fuel: The Science and Technology of Fuel and Energy, 175, 157-163 (2016)
Guangyi Li et al.
ChemSusChem, 5(10), 1958-1966 (2012-08-22)
Hydroxyalkylation-alkylation (HAA) coupled with hydrodeoxygenation is a promising route for the synthesis of renewable high-quality diesel or jet fuel. In this work, a series of solid-acid catalysts were firstly used for HAA between lignocellulose-derived furan and carbonyl compounds. Among the
V Ravindranath et al.
Toxicology and applied pharmacology, 85(1), 78-91 (1986-08-01)
Administration of a single ip dose of 2-methylfuran (2-MF) to male Sprague-Dawley rats at a dose of 100 mg/kg produced centrilobular necrosis of the liver and bronchial injury of the lung, the severity of the lesions increasing with increasing doses
Renewable production of phthalic anhydride from biomass-derived furan and maleic anhydride.
Mahmoud E, et al.
Green Chemistry, 16(1), 167-175 (2014)
Jinglei Cui et al.
ChemSusChem, 9(11), 1259-1262 (2016-04-28)
An efficient process was designed for the synthesis of furfuryl alcohol and 2-methylfuran from xylose using a continuous fixed-bed reactor over a catalyst combining Hβ zeolite and Cu/ZnO/Al2 O3 in γ-butyrolactone (GBL)/water as solvent. The cooperative effect of Hβ zeolite

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service