Skip to Content
Merck
All Photos(1)

Key Documents

HAM203349

Hamilton® chromatography syringe

for CTC/LEAP PAL LC Autosampler, model, 1750 FN CTC, fixed needle, volume × L 500 μL × 51 mm, needle gauge 22

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122001
NACRES:
NB.03

material

Gastight syringe

grade

for chromatography

feature

autoclavable: no
model 1750 FN CTC, fixed needle

manufacturer/tradename

Hamilton

parameter

20-50 °C temp. range (68-122 °F)
700 psig pressure

needle I.D. × O.D.

0.41 mm × 0.72 mm

needle gauge

22

volume × L

500 μL × 51 mm

Looking for similar products? Visit Product Comparison Guide

General description

S-Line syringes deliver exceptional performance for everyday use in HPLC autosampler injections at a great value. The C-Line syringe minimizes the potential for bent needles with a keyed flange design. X-Type syringes are designed for sensitive LC samples (i.e. proteins, peptides, phospholipids, smaller amines, etc.) with a specially coated needle and inner syringe barrel to minimize sample adsorption.

Legal Information

Hamilton is a registered trademark of Hamilton Co.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service