Skip to Content
Merck
All Photos(2)

Documents

H20601

Sigma-Aldrich

3-Hydroxybenzyl alcohol

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H4CH2OH
CAS Number:
Molecular Weight:
124.14
Beilstein:
2041500
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

mp

69-72 °C (lit.)

SMILES string

OCc1cccc(O)c1

InChI

1S/C7H8O2/c8-5-6-2-1-3-7(9)4-6/h1-4,8-9H,5H2

InChI key

OKVJCVWFVRATSG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Bruze et al.
Acta dermato-venereologica, 65(6), 548-551 (1985-01-01)
Thirteen patients with contact allergy to phenol-formaldehyde resins (P-F-R) were patch tested with 3-methylol phenol, 2,4-dimethylol phenol and 2,6-dimethylol phenol. Nine patients reacted to at least 1 compound, all giving positive test responses to 2,4-dimethylol phenol. Seven patients reacted simultaneously
Cristina Alfaro et al.
Mutation research, 539(1-2), 187-194 (2003-09-02)
In the search for new natural products with anti-oxidant activity, we have combined the cell-free assay based on the scavenging of the stable radical 2,2-diphenyl-1-picrylhydrazyl (DPPH), with a bioassay that detects oxidative mutagens. This bioassay uses a new Escherichia coli
L G Fenoll et al.
Biological chemistry, 381(4), 313-320 (2000-06-06)
The relationship between the structure and activity of meta- and para-hydroxylated monophenols was studied during their tyrosinase-catalysed hydroxylation and the rate-limiting steps of the reaction mechanism were identified. The para-hydroxylated substrates permit us to study the effect of a substituent
C Ganesh Kumar et al.
Journal of applied microbiology, 122(6), 1518-1528 (2017-03-18)
The aim of the study was to purify and characterize a bioactive compound from Aspergillus nidulans strain KZR-132 and its biological evaluation. A bioactive extolite was purified from A. nidulans strain KZR-132, and its chemical structure was elucidated as 3-hydroxylbenzyl
Sophie N B Selby-Pham et al.
Food & function, 11(1), 907-920 (2020-01-17)
After oil extraction, palm fruit biomass contains abundant water-soluble phytochemicals (PCs) with proven bioactivity in regulating oxidative stress and inflammation (OSI). For optimal bioefficacy following oral consumption, the pharmacokinetic plasma peak (Tmax) should be bio-matched with the onset of OSI

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service