Skip to Content
Merck
All Photos(2)

Key Documents

D7600

Sigma-Aldrich

1,2:5,6-Di-O-isopropylidene-α-D-glucofuranose

98%

Synonym(s):

1,2,5,6-diisopropylidene-D-glucose, D-Glucose diacetonide, Diacetone-D-glucose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H20O6
CAS Number:
Molecular Weight:
260.28
Beilstein:
84386
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

powder

optical activity

[α]20/D −18°, c = 1% in H2O

mp

109-113 °C (lit.)

SMILES string

[H][C@@]1(O[C@H]2O[C@@H](O[C@@H]2[C@H]1O)C(Cl)(Cl)Cl)[C@H]3COC(C)(C)O3

InChI

1S/C12H20O6/c1-11(2)14-5-6(16-11)8-7(13)9-10(15-8)18-12(3,4)17-9/h6-10,13H,5H2,1-4H3/t6-,7+,8-,9-,10-/m1/s1

InChI key

KEJGAYKWRDILTF-JDDHQFAOSA-N

Application

1,2:5,6-Di-O-isopropylidene-α-D-glucofuranose can be used as a starting material to prepare:
  • Biologically active L-acovenose, 6-deoxy-L-idose and, carbanucleoside enantiomers.
  • Vinyl ether-based chiral carbohydrate synthon by reacting with acetylene using superbase catalytic systems.
  • Fluoro-thiofuranosyl nucleosides of biological importance.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Direct vinylation of glucose derivatives with acetylene
Trofimov BA, et al.
Tetrahedron, 63(47), 11661-11665 (2007)
An efficient synthesis of 3-fluoro-5-thio-xylofuranosyl nucleosides of thymine, uracil, and 5-fluorouracil as potential antitumor or/and antiviral agents
Tsoukala E, et al.
Bioorganic & Medicinal Chemistry, 15(9), 3241-3247 (2007)
Guylaine M Defossemont et al.
Carbohydrate research, 338(6), 563-565 (2003-04-02)
The synthesis and characterisation of a novel chiral bicyclic oxacaprolactone is reported. The choice of diisopropylidene-D-glucose as a starting material allowed selective introduction of the synthetic equivalent necessary for the formation of the seven-membered ring of the lactone, i.e., one
S C Hung et al.
Carbohydrate research, 331(4), 369-374 (2001-06-12)
A practical route toward the synthesis of 6-deoxy-L-idose and L-acovenose from 1,2:5,6-di-O-isopropylidene-alpha-D-glucofuranose is described. Key steps include the stereoselective hydrogenation of 6-deoxy-1,2:3,5-di-O-isopropylidene-alpha-D-xylo-hex-5-enofuranose, regioselective protection of 6-deoxy-1,2-O-isopropylidene-beta-L-idofuranose at 0-5, and epimerisation of 6-deoxy-5-O-tert-butyldimethylsilyl-1,2-O-isopropylidene-beta-L-idofuranose at C-3.
T Z Csáky
Journal of medicine, 16(5-6), 575-586 (1985-01-01)
Based on a previous observation, it was postulated that dimethylsulfoxide (DMSO) acts as a carrier-model in the intestinal absorption of glucose and galactose (Csáky and Ho, 1966). It was further hypothetized that DMSO forms a loosely-bonded hydrophobic complex with the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service