Skip to Content
Merck
All Photos(1)

Key Documents

CDS020790

Sigma-Aldrich

Xanthosine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12N4O6
Molecular Weight:
284.23
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

description

AldrichCPR

form

solid

SMILES string

O[C@H]1[C@@H](O)[C@H](N(C=N2)C3=C2C(NC(N3)=O)=O)O[C@@H]1CO

InChI

1S/C10H12N4O6/c15-1-3-5(16)6(17)9(20-3)14-2-11-4-7(14)12-10(19)13-8(4)18/h2-3,5-6,9,15-17H,1H2,(H2,12,13,18,19)/t3-,5-,6-,9-/m1/s1

InChI key

UBORTCNDUKBEOP-UUOKFMHZSA-N

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY, (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE, OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY, WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Yongzhao Guo et al.
Environmental science & technology, 51(16), 9155-9164 (2017-07-25)
Substrate concentrations generally fluctuate in wastewaters. However, how anammox biomass behaves to overcome the stress of alternating feed loadings remains unclear. Here, we combined long-term reactor operation, batch tests, 16S rRNA transcript sequencing, and metabolomics analysis to investigate the aggregation
Ratan K Choudhary et al.
BMC cell biology, 13, 14-14 (2012-06-16)
Mammary stem cells are critical for growth and maintenance of the mammary gland and therefore are of considerable interest for improving productivity and efficiency of dairy animals. Xanthosine treatment has been demonstrated to promote expansion of putative mammary stem cells
Guangguo Tan et al.
PloS one, 7(4), e34157-e34157 (2012-04-12)
In spite of great advances in target-oriented Western medicine for treating myocardial infarction (MI), it is still a leading cause of death in a worldwide epidemic. In contrast to Western medicine, Traditional Chinese medicine (TCM) uses a holistic and synergistic
Yue-Hong Zhang et al.
Nature communications, 11(1), 1473-1473 (2020-03-21)
Caffeine is a major component of xanthine alkaloids and commonly consumed in many popular beverages. Due to its occasional side effects, reduction of caffeine in a natural way is of great importance and economic significance. Recent studies reveal that caffeine
Rouzbeh R Taghizadeh et al.
Journal of biomedicine & biotechnology, 2008, 453590-453590 (2008-04-11)
The stable expression of reporter genes in adult stem cells (ASCs) has important applications in stem cell biology. The ability to integrate a noncytotoxic, fluorescent reporter gene into the genome of ASCs with the capability to track ASCs and their

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service