Skip to Content
Merck
All Photos(1)

Documents

C52276

Sigma-Aldrich

4-Phenylbenzyl chloride

98%

Synonym(s):

4-(Chloromethyl)biphenyl

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5C6H4CH2Cl
CAS Number:
Molecular Weight:
202.68
Beilstein:
1863327
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

powder

mp

70-73 °C (lit.)

SMILES string

ClCc1ccc(cc1)-c2ccccc2

InChI

1S/C13H11Cl/c14-10-11-6-8-13(9-7-11)12-4-2-1-3-5-12/h1-9H,10H2

InChI key

HLQZCRVEEQKNMS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Is 4CMB clastogenic in vivo? Preliminary results from the mouse transplacental micronucleus test.
R J Cole et al.
Mutation research, 100(1-4), 379-380 (1982-01-01)
Activity of 4CMB in the mouse-skin sebaceous-gland suppression test.
J Ashby et al.
Mutation research, 100(1-4), 391-393 (1982-01-01)
An evaluation of the mutagenic potential of 4-chloromethylbiphenyl (4CMB) using the micronucleus test.
J C Richardson et al.
Mutation research, 100(1-4), 375-378 (1982-01-01)
Activity of 4-chloromethylbiphenyl.
J Ashby et al.
Mutation research, 135(1), 51-52 (1984-01-01)
R Mirzayans et al.
Mutation research, 145(3), 209-215 (1985-05-01)
Benzyl chloride (BC) and 4-chloromethylbiphenyl (4CMB) induce a class of alkaline-stable DNA damage in human cells which, like UV-induced pyrimidine dimers, undergoes repair at a slow rate by an excision-repair pathway which can be inhibited by cytosine arabinoside (araC). In

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service